Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 62664753-62665010 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLCO4A1
Entrez Gene ID
28231
Gene Name
solute carrier organic anion transporter family member 4A1
Gene Aliases
OATP-E, OATP1, OATP4A1, OATPE, OATPRP1, POAT, SLC21A12
UniGene
Hs.235782
Gene Chromosome Location
Chr.20: 62642445-62690700 on Build GRCh38
Transcripts
NM_016354.3 XM_005260203.2 XM_011528792.2 XM_017027826.1 XM_017027827.1 NR_024470.1

Gene Symbol
SLCO4A1-AS1
Entrez Gene ID
100127888
Gene Name
SLCO4A1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.671273
Gene Chromosome Location
Chr.20: 62663027-62666621 on Build GRCh38
Transcripts
NM_016354.3 XM_005260203.2 XM_011528792.2 XM_017027826.1 XM_017027827.1 NR_024470.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7182307 c.1122-1G>A p.? Chr.20: 62664933-62664933 on Build GRCh38
6783954 c.1122-6G>A p.? Chr.20: 62664928-62664928 on Build GRCh38
7390876 c.1131G>C p.W377C Chr.20: 62664943-62664943 on Build GRCh38
7153590 c.1164C>G p.L388L Chr.20: 62664976-62664976 on Build GRCh38
5482914 c.1174G>A p.G392R Chr.20: 62664986-62664986 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370641386 Chr.20: 62664907-62664907 on Build GRCh38
rs1020565651 Chr.20: 62664781-62664784 on Build GRCh38
rs563579653 Chr.20: 62664844-62664844 on Build GRCh38
rs775077289 Chr.20: 62664989-62664989 on Build GRCh38
rs1057360352 Chr.20: 62664959-62664959 on Build GRCh38
rs188188055 Chr.20: 62664876-62664876 on Build GRCh38
rs1001107108 Chr.20: 62664809-62664809 on Build GRCh38
rs763593918 Chr.20: 62664886-62664886 on Build GRCh38
rs771342944 Chr.20: 62664978-62664978 on Build GRCh38
rs746347335 Chr.20: 62664825-62664825 on Build GRCh38
rs878983275 Chr.20: 62664847-62664847 on Build GRCh38
rs973265673 Chr.20: 62664929-62664929 on Build GRCh38
rs546283820 Chr.20: 62664911-62664911 on Build GRCh38
rs897368064 Chr.20: 62664879-62664879 on Build GRCh38
rs968523839 Chr.20: 62664786-62664786 on Build GRCh38
rs570220517 Chr.20: 62664885-62664885 on Build GRCh38
rs192519907 Chr.20: 62664905-62664905 on Build GRCh38
rs1048509681 Chr.20: 62664898-62664898 on Build GRCh38
rs988716877 Chr.20: 62664773-62664773 on Build GRCh38
rs760130941 Chr.20: 62664985-62664985 on Build GRCh38
rs373862291 Chr.20: 62664964-62664964 on Build GRCh38
rs183755475 Chr.20: 62664955-62664955 on Build GRCh38
rs925352353 Chr.20: 62664800-62664800 on Build GRCh38
rs774678126 Chr.20: 62664983-62664983 on Build GRCh38
rs756404185 Chr.20: 62664927-62664927 on Build GRCh38
rs758235087 Chr.20: 62664942-62664942 on Build GRCh38
rs866419194 Chr.20: 62664984-62664984 on Build GRCh38
rs749511648 Chr.20: 62664967-62664967 on Build GRCh38
rs781360831 Chr.20: 62664953-62664953 on Build GRCh38
rs2277774 Chr.20: 62664916-62664916 on Build GRCh38
rs554990110 Chr.20: 62664937-62664937 on Build GRCh38
rs975237553 Chr.20: 62664870-62664870 on Build GRCh38
rs750497564 Chr.20: 62664916-62664917 on Build GRCh38
rs902792508 Chr.20: 62664808-62664808 on Build GRCh38
rs530446705 Chr.20: 62664875-62664875 on Build GRCh38
rs964852014 Chr.20: 62664853-62664853 on Build GRCh38
rs1001217622 Chr.20: 62664913-62664913 on Build GRCh38
rs979553877 Chr.20: 62664796-62664796 on Build GRCh38
rs112024334 Chr.20: 62664923-62664923 on Build GRCh38
rs371331068 Chr.20: 62664963-62664963 on Build GRCh38
rs954948435 Chr.20: 62664889-62664889 on Build GRCh38
rs778834099 Chr.20: 62664933-62664933 on Build GRCh38
rs944314941 Chr.20: 62664841-62664841 on Build GRCh38
rs113273141 Chr.20: 62664831-62664831 on Build GRCh38

View Full Product Details