Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 36626194-36626433 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF382
Entrez Gene ID
84911
Gene Name
zinc finger protein 382
Gene Aliases
KS1
UniGene
Hs.631591
Gene Chromosome Location
Chr.19: 36605305-36628597 on Build GRCh38
Transcripts
NM_001256838.1 NM_032825.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6829720 c.480G>T p.K160N Chr.19: 36626377-36626377 on Build GRCh38
6829723 c.494G>A p.G165E Chr.19: 36626394-36626394 on Build GRCh38
4077401 c.323T>G p.L108R Chr.19: 36626223-36626223 on Build GRCh38
5144815 c.372G>T p.K124N Chr.19: 36626272-36626272 on Build GRCh38
6879984 c.421A>T p.K141* Chr.19: 36626318-36626318 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs3108171 Chr.19: 36626400-36626400 on Build GRCh38
rs772050971 Chr.19: 36626234-36626234 on Build GRCh38
rs781264235 Chr.19: 36626386-36626386 on Build GRCh38
rs778805731 Chr.19: 36626222-36626222 on Build GRCh38
rs763389271 Chr.19: 36626263-36626263 on Build GRCh38
rs754114549 Chr.19: 36626348-36626348 on Build GRCh38
rs747949757 Chr.19: 36626361-36626361 on Build GRCh38
rs142570474 Chr.19: 36626277-36626277 on Build GRCh38
rs776464213 Chr.19: 36626262-36626262 on Build GRCh38
rs764611801 Chr.19: 36626276-36626276 on Build GRCh38
rs1032904837 Chr.19: 36626252-36626252 on Build GRCh38
rs768086756 Chr.19: 36626288-36626288 on Build GRCh38
rs778609516 Chr.19: 36626353-36626353 on Build GRCh38
rs772538250 Chr.19: 36626235-36626235 on Build GRCh38
rs777795980 Chr.19: 36626376-36626376 on Build GRCh38
rs770449856 Chr.19: 36626260-36626260 on Build GRCh38
rs533817629 Chr.19: 36626351-36626351 on Build GRCh38
rs760253473 Chr.19: 36626244-36626244 on Build GRCh38
rs552027724 Chr.19: 36626341-36626341 on Build GRCh38
rs757262971 Chr.19: 36626311-36626324 on Build GRCh38
rs750423107 Chr.19: 36626332-36626332 on Build GRCh38
rs766485216 Chr.19: 36626342-36626342 on Build GRCh38
rs748147772 Chr.19: 36626233-36626233 on Build GRCh38
rs758219017 Chr.19: 36626366-36626366 on Build GRCh38
rs566322984 Chr.19: 36626339-36626339 on Build GRCh38
rs762347609 Chr.19: 36626279-36626279 on Build GRCh38
rs747266278 Chr.19: 36626228-36626228 on Build GRCh38
rs746468500 Chr.19: 36626384-36626384 on Build GRCh38
rs181678050 Chr.19: 36626393-36626393 on Build GRCh38

View Full Product Details