Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 17501383-17501624 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USH1C
Entrez Gene ID
10083
Gene Name
USH1 protein network component harmonin
Gene Aliases
AIE-75, DFNB18, DFNB18A, NY-CO-37, NY-CO-38, PDZ-45, PDZ-73, PDZ-73/NY-CO-38, PDZ73, PDZD7C, ush1cpst
UniGene
Hs.502072
Gene Chromosome Location
Chr.11: 17493895-17544416 on Build GRCh38
Transcripts
NM_001297764.1 NM_005709.3 NM_153676.3 NR_123738.1 XM_011519832.2 XM_017017075.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4535931 c.1350G>A p.G450G Chr.11: 17501512-17501512 on Build GRCh38
5541436 c.2233C>T p.P745S Chr.11: 17501529-17501529 on Build GRCh38
5541437 c.1333C>T p.P445S Chr.11: 17501529-17501529 on Build GRCh38
1231911 c.2260C>T p.R754W Chr.11: 17501502-17501502 on Build GRCh38
324196 c.2227-2A>G p.? Chr.11: 17501537-17501537 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745722516 Chr.11: 17501482-17501484 on Build GRCh38
rs757149073 Chr.11: 17501473-17501473 on Build GRCh38
rs201600193 Chr.11: 17501522-17501522 on Build GRCh38
rs758112290 Chr.11: 17501543-17501543 on Build GRCh38
rs766976437 Chr.11: 17501508-17501508 on Build GRCh38
rs778799555 Chr.11: 17501475-17501475 on Build GRCh38
rs779277441 Chr.11: 17501561-17501561 on Build GRCh38
rs778110397 Chr.11: 17501536-17501536 on Build GRCh38
rs753703742 Chr.11: 17501492-17501492 on Build GRCh38
rs758893475 Chr.11: 17501452-17501452 on Build GRCh38
rs780634496 Chr.11: 17501581-17501581 on Build GRCh38
rs757163581 Chr.11: 17501493-17501493 on Build GRCh38
rs776356060 Chr.11: 17501476-17501476 on Build GRCh38
rs566698194 Chr.11: 17501548-17501548 on Build GRCh38
rs151251262 Chr.11: 17501497-17501497 on Build GRCh38
rs369119582 Chr.11: 17501414-17501414 on Build GRCh38
rs376293679 Chr.11: 17501461-17501461 on Build GRCh38
rs775252771 Chr.11: 17501560-17501561 on Build GRCh38
rs374196365 Chr.11: 17501441-17501441 on Build GRCh38
rs71484772 Chr.11: 17501601-17501601 on Build GRCh38
rs186936300 Chr.11: 17501430-17501430 on Build GRCh38
rs373271463 Chr.11: 17501564-17501564 on Build GRCh38
rs143778195 Chr.11: 17501483-17501483 on Build GRCh38
rs746911298 Chr.11: 17501445-17501445 on Build GRCh38
rs143803480 Chr.11: 17501502-17501502 on Build GRCh38
rs769480279 Chr.11: 17501585-17501585 on Build GRCh38
rs548090508 Chr.11: 17501541-17501541 on Build GRCh38
rs756079615 Chr.11: 17501410-17501410 on Build GRCh38
rs980897792 Chr.11: 17501467-17501467 on Build GRCh38
rs886048059 Chr.11: 17501467-17501467 on Build GRCh38
rs199537540 Chr.11: 17501460-17501460 on Build GRCh38
rs145832659 Chr.11: 17501490-17501490 on Build GRCh38
rs974123483 Chr.11: 17501519-17501519 on Build GRCh38
rs569102513 Chr.11: 17501462-17501463 on Build GRCh38
rs767662810 Chr.11: 17501517-17501517 on Build GRCh38
rs367888870 Chr.11: 17501584-17501584 on Build GRCh38
rs891062796 Chr.11: 17501587-17501587 on Build GRCh38
rs750962113 Chr.11: 17501449-17501449 on Build GRCh38
rs770171347 Chr.11: 17501501-17501501 on Build GRCh38
rs751998841 Chr.11: 17501509-17501509 on Build GRCh38
rs769331847 Chr.11: 17501554-17501554 on Build GRCh38
rs1028903986 Chr.11: 17501426-17501426 on Build GRCh38
rs114971046 Chr.11: 17501453-17501453 on Build GRCh38
rs370437884 Chr.11: 17501516-17501516 on Build GRCh38
rs765522476 Chr.11: 17501447-17501447 on Build GRCh38
rs1031731022 Chr.11: 17501444-17501444 on Build GRCh38
rs376011428 Chr.11: 17501570-17501570 on Build GRCh38
rs201211922 Chr.11: 17501574-17501574 on Build GRCh38
rs1004622706 Chr.11: 17501539-17501539 on Build GRCh38
rs1050856232 Chr.11: 17501580-17501580 on Build GRCh38
rs370092421 Chr.11: 17501459-17501459 on Build GRCh38
rs756452832 Chr.11: 17501524-17501524 on Build GRCh38

View Full Product Details