Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 36393875-36394144 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFP82
Entrez Gene ID
284406
Gene Name
ZFP82 zinc finger protein
Gene Aliases
ZNF545
UniGene
Hs.558734
Gene Chromosome Location
Chr.19: 36383102-36418674 on Build GRCh38
Transcripts
NM_001321917.1 NM_001321918.1 NM_001321919.1 NM_133466.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6524912 c.332G>T p.G111V Chr.19: 36394008-36394008 on Build GRCh38
4077371 c.380T>C p.I127T Chr.19: 36393960-36393960 on Build GRCh38
1393074 c.275A>G p.D92G Chr.19: 36394065-36394065 on Build GRCh38
3532926 c.407G>A p.G136E Chr.19: 36393933-36393933 on Build GRCh38
1393073 c.358G>T p.D120Y Chr.19: 36393982-36393982 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144032300 Chr.19: 36393979-36393979 on Build GRCh38
rs148540683 Chr.19: 36393982-36393982 on Build GRCh38
rs764982529 Chr.19: 36394099-36394099 on Build GRCh38
rs963573736 Chr.19: 36393935-36393935 on Build GRCh38
rs945254327 Chr.19: 36394084-36394084 on Build GRCh38
rs756799775 Chr.19: 36393986-36393986 on Build GRCh38
rs370541972 Chr.19: 36394052-36394052 on Build GRCh38
rs773420403 Chr.19: 36394050-36394050 on Build GRCh38
rs748645640 Chr.19: 36394011-36394011 on Build GRCh38
rs750725834 Chr.19: 36393960-36393961 on Build GRCh38
rs752586717 Chr.19: 36393931-36393931 on Build GRCh38
rs926359846 Chr.19: 36393994-36393994 on Build GRCh38
rs1028168703 Chr.19: 36394059-36394059 on Build GRCh38
rs779462448 Chr.19: 36394009-36394009 on Build GRCh38
rs776665195 Chr.19: 36394095-36394095 on Build GRCh38
rs1016802799 Chr.19: 36393922-36393922 on Build GRCh38
rs780655678 Chr.19: 36394005-36394005 on Build GRCh38
rs950270480 Chr.19: 36393959-36393959 on Build GRCh38
rs745557666 Chr.19: 36394007-36394007 on Build GRCh38
rs765098015 Chr.19: 36393896-36393896 on Build GRCh38
rs759830950 Chr.19: 36394097-36394097 on Build GRCh38
rs573960775 Chr.19: 36394015-36394015 on Build GRCh38
rs775310362 Chr.19: 36394115-36394115 on Build GRCh38
rs561996621 Chr.19: 36393960-36393960 on Build GRCh38
rs35344339 Chr.19: 36393903-36393904 on Build GRCh38
rs375155109 Chr.19: 36394063-36394063 on Build GRCh38

View Full Product Details