Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89691356-89691629 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK10
Entrez Gene ID
8558
Gene Name
cyclin dependent kinase 10
Gene Aliases
PISSLRE
UniGene
Hs.699177
Gene Chromosome Location
Chr.16: 89686612-89696364 on Build GRCh38
Transcripts
NM_001098533.2 NM_001160367.1 NM_052987.3 NM_052988.4 NR_027702.1 NR_027703.1 XM_006721308.2 XM_006721310.2 XM_011523405.2 XM_011523406.2 XM_011523407.2 XM_011523408.2 XM_011523410.2 XM_011523411.2 XM_011523412.1 XM_011523413.2 XM_011523414.2 XM_011523415.2 XM_011523416.2 XM_011523417.2 XM_017023806.1 XM_017023807.1 XM_017023808.1 XM_017023809.1 XM_017023810.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7307217 c.254G>A p.R85Q Chr.16: 89691464-89691464 on Build GRCh38
1247906 c.335G>C p.S112T Chr.16: 89691545-89691545 on Build GRCh38
974920 c.280C>T p.R94C Chr.16: 89691490-89691490 on Build GRCh38
7110119 c.81C>T p.I27I Chr.16: 89691504-89691504 on Build GRCh38
4671066 c.275G>A p.R92H Chr.16: 89691485-89691485 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs538408587 Chr.16: 89691382-89691382 on Build GRCh38
rs938269727 Chr.16: 89691522-89691522 on Build GRCh38
rs1055365150 Chr.16: 89691534-89691534 on Build GRCh38
rs770683138 Chr.16: 89691582-89691582 on Build GRCh38
rs757830367 Chr.16: 89691417-89691417 on Build GRCh38
rs778233593 Chr.16: 89691408-89691408 on Build GRCh38
rs754530842 Chr.16: 89691557-89691557 on Build GRCh38
rs370885215 Chr.16: 89691484-89691484 on Build GRCh38
rs757712230 Chr.16: 89691464-89691464 on Build GRCh38
rs759646229 Chr.16: 89691447-89691447 on Build GRCh38
rs570732324 Chr.16: 89691570-89691570 on Build GRCh38
rs753023971 Chr.16: 89691402-89691402 on Build GRCh38
rs950312171 Chr.16: 89691394-89691394 on Build GRCh38
rs184050237 Chr.16: 89691490-89691490 on Build GRCh38
rs191532111 Chr.16: 89691376-89691376 on Build GRCh38
rs376178897 Chr.16: 89691569-89691569 on Build GRCh38
rs746477654 Chr.16: 89691474-89691474 on Build GRCh38
rs769750795 Chr.16: 89691428-89691428 on Build GRCh38
rs751005431 Chr.16: 89691553-89691553 on Build GRCh38
rs779556993 Chr.16: 89691419-89691419 on Build GRCh38
rs562096833 Chr.16: 89691436-89691436 on Build GRCh38
rs755762652 Chr.16: 89691565-89691565 on Build GRCh38
rs556882238 Chr.16: 89691398-89691398 on Build GRCh38
rs370747969 Chr.16: 89691549-89691549 on Build GRCh38
rs138561644 Chr.16: 89691473-89691473 on Build GRCh38
rs576480961 Chr.16: 89691420-89691420 on Build GRCh38
rs373968493 Chr.16: 89691485-89691485 on Build GRCh38
rs770863327 Chr.16: 89691494-89691494 on Build GRCh38
rs543590749 Chr.16: 89691434-89691434 on Build GRCh38
rs747932338 Chr.16: 89691426-89691426 on Build GRCh38
rs780215532 Chr.16: 89691473-89691483 on Build GRCh38
rs778882774 Chr.16: 89691586-89691586 on Build GRCh38
rs754584020 Chr.16: 89691475-89691475 on Build GRCh38
rs981622155 Chr.16: 89691592-89691592 on Build GRCh38
rs749041801 Chr.16: 89691491-89691491 on Build GRCh38
rs747098145 Chr.16: 89691565-89691568 on Build GRCh38
rs765750522 Chr.16: 89691550-89691550 on Build GRCh38
rs776445439 Chr.16: 89691422-89691422 on Build GRCh38
rs764224553 Chr.16: 89691528-89691528 on Build GRCh38
rs887182112 Chr.16: 89691395-89691395 on Build GRCh38
rs767682288 Chr.16: 89691453-89691453 on Build GRCh38
rs55819627 Chr.16: 89691497-89691497 on Build GRCh38
rs756442193 Chr.16: 89691459-89691459 on Build GRCh38
rs372926316 Chr.16: 89691416-89691416 on Build GRCh38
rs752968620 Chr.16: 89691457-89691457 on Build GRCh38
rs144667632 Chr.16: 89691463-89691463 on Build GRCh38
rs926917993 Chr.16: 89691385-89691385 on Build GRCh38
rs533676978 Chr.16: 89691561-89691561 on Build GRCh38
rs748444226 Chr.16: 89691464-89691464 on Build GRCh38
rs913516277 Chr.16: 89691386-89691386 on Build GRCh38
rs1028901469 Chr.16: 89691430-89691430 on Build GRCh38
rs377332718 Chr.16: 89691421-89691421 on Build GRCh38
rs780908690 Chr.16: 89691560-89691560 on Build GRCh38
rs780959674 Chr.16: 89691476-89691476 on Build GRCh38
rs754188486 Chr.16: 89691531-89691531 on Build GRCh38
rs188735443 Chr.16: 89691504-89691504 on Build GRCh38
rs749065322 Chr.16: 89691573-89691573 on Build GRCh38
rs766241442 Chr.16: 89691439-89691439 on Build GRCh38
rs528022728 Chr.16: 89691435-89691435 on Build GRCh38
rs375735637 Chr.16: 89691406-89691406 on Build GRCh38
rs772182758 Chr.16: 89691498-89691498 on Build GRCh38
rs967091595 Chr.16: 89691462-89691462 on Build GRCh38
rs774423581 Chr.16: 89691441-89691441 on Build GRCh38

View Full Product Details