Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 145859686-145859959 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRXCR2
Entrez Gene ID
643226
Gene Name
glutaredoxin and cysteine rich domain containing 2
Gene Aliases
DFNB101
UniGene
Hs.570895
Gene Chromosome Location
Chr.5: 145858538-145931673 on Build GRCh38
Transcripts
NM_001080516.1 XM_017009708.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
420849 c.690G>A p.L230L Chr.5: 145859790-145859790 on Build GRCh38
6169714 c.604G>T p.G202W Chr.5: 145859876-145859876 on Build GRCh38
1739169 c.704G>T p.C235F Chr.5: 145859776-145859776 on Build GRCh38
6169713 c.568_569GG>TT p.G190L Chr.5: 145859911-145859912 on Build GRCh38
5723789 c.626C>T p.S209F Chr.5: 145859854-145859854 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376451689 Chr.5: 145859716-145859716 on Build GRCh38
rs770653814 Chr.5: 145859854-145859854 on Build GRCh38
rs745650659 Chr.5: 145859912-145859912 on Build GRCh38
rs776961131 Chr.5: 145859776-145859776 on Build GRCh38
rs772716837 Chr.5: 145859731-145859754 on Build GRCh38
rs778713809 Chr.5: 145859860-145859860 on Build GRCh38
rs762204674 Chr.5: 145859823-145859823 on Build GRCh38
rs768725056 Chr.5: 145859771-145859771 on Build GRCh38
rs759001784 Chr.5: 145859829-145859829 on Build GRCh38
rs576525485 Chr.5: 145859817-145859817 on Build GRCh38
rs200294198 Chr.5: 145859800-145859800 on Build GRCh38
rs757758226 Chr.5: 145859738-145859738 on Build GRCh38
rs866343537 Chr.5: 145859807-145859808 on Build GRCh38
rs750908567 Chr.5: 145859827-145859827 on Build GRCh38
rs759494548 Chr.5: 145859798-145859798 on Build GRCh38
rs763420784 Chr.5: 145859880-145859880 on Build GRCh38
rs374291185 Chr.5: 145859797-145859797 on Build GRCh38
rs587777424 Chr.5: 145859766-145859767 on Build GRCh38
rs779793231 Chr.5: 145859925-145859925 on Build GRCh38
rs760103412 Chr.5: 145859808-145859809 on Build GRCh38
rs760282045 Chr.5: 145859804-145859804 on Build GRCh38
rs535371236 Chr.5: 145859825-145859825 on Build GRCh38
rs746384864 Chr.5: 145859751-145859751 on Build GRCh38
rs755755532 Chr.5: 145859729-145859729 on Build GRCh38
rs770535368 Chr.5: 145859848-145859849 on Build GRCh38
rs201767665 Chr.5: 145859927-145859927 on Build GRCh38
rs752878636 Chr.5: 145859892-145859892 on Build GRCh38
rs765655672 Chr.5: 145859879-145859879 on Build GRCh38
rs752347075 Chr.5: 145859723-145859723 on Build GRCh38
rs768005413 Chr.5: 145859929-145859929 on Build GRCh38
rs139847108 Chr.5: 145859871-145859871 on Build GRCh38
rs761577147 Chr.5: 145859878-145859878 on Build GRCh38
rs781344559 Chr.5: 145859891-145859891 on Build GRCh38
rs201508591 Chr.5: 145859736-145859736 on Build GRCh38
rs367826984 Chr.5: 145859900-145859900 on Build GRCh38
rs192698368 Chr.5: 145859853-145859853 on Build GRCh38
rs761960449 Chr.5: 145859730-145859730 on Build GRCh38
rs1040501725 Chr.5: 145859883-145859883 on Build GRCh38
rs199737180 Chr.5: 145859843-145859843 on Build GRCh38
rs779560934 Chr.5: 145859741-145859741 on Build GRCh38
rs994990403 Chr.5: 145859780-145859780 on Build GRCh38
rs752175323 Chr.5: 145859890-145859890 on Build GRCh38
rs747211639 Chr.5: 145859770-145859770 on Build GRCh38
rs376485351 Chr.5: 145859732-145859732 on Build GRCh38
rs576894412 Chr.5: 145859931-145859931 on Build GRCh38
rs780016246 Chr.5: 145859769-145859769 on Build GRCh38
rs115520168 Chr.5: 145859901-145859901 on Build GRCh38
rs745704053 Chr.5: 145859864-145859864 on Build GRCh38
rs771973384 Chr.5: 145859913-145859913 on Build GRCh38
rs970602411 Chr.5: 145859743-145859743 on Build GRCh38
rs781265525 Chr.5: 145859844-145859844 on Build GRCh38
rs758492472 Chr.5: 145859762-145859762 on Build GRCh38
rs952270962 Chr.5: 145859830-145859830 on Build GRCh38
rs751566675 Chr.5: 145859841-145859841 on Build GRCh38
rs922041995 Chr.5: 145859870-145859870 on Build GRCh38
rs763644996 Chr.5: 145859813-145859813 on Build GRCh38
rs72816449 Chr.5: 145859720-145859720 on Build GRCh38
rs770778426 Chr.5: 145859792-145859792 on Build GRCh38
rs747698755 Chr.5: 145859934-145859934 on Build GRCh38
rs774832434 Chr.5: 145859801-145859801 on Build GRCh38
rs749067935 Chr.5: 145859784-145859784 on Build GRCh38
rs760103355 Chr.5: 145859872-145859872 on Build GRCh38
rs766781755 Chr.5: 145859882-145859882 on Build GRCh38

View Full Product Details