Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 74038085-74038350 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXCL3
Entrez Gene ID
2921
Gene Name
C-X-C motif chemokine ligand 3
Gene Aliases
CINC-2b, GRO3, GROg, MIP-2b, MIP2B, SCYB3
UniGene
Hs.89690
Gene Chromosome Location
Chr.4: 74036589-74038773 on Build GRCh38
Transcripts
NM_002090.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1202736 c.300A>G p.I100M Chr.4: 74038101-74038101 on Build GRCh38
4845297 c.282G>A p.Q94Q Chr.4: 74038119-74038119 on Build GRCh38
4675706 c.190T>C p.S64P Chr.4: 74038324-74038324 on Build GRCh38
1202735 c.306C>A p.N102K Chr.4: 74038095-74038095 on Build GRCh38
3605771 c.292G>A p.E98K Chr.4: 74038109-74038109 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs927335185 Chr.4: 74038159-74038159 on Build GRCh38
rs755866833 Chr.4: 74038209-74038209 on Build GRCh38
rs752940632 Chr.4: 74038275-74038275 on Build GRCh38
rs574512340 Chr.4: 74038185-74038185 on Build GRCh38
rs763680466 Chr.4: 74038154-74038154 on Build GRCh38
rs561019043 Chr.4: 74038280-74038280 on Build GRCh38
rs766328028 Chr.4: 74038273-74038273 on Build GRCh38
rs556351004 Chr.4: 74038203-74038203 on Build GRCh38
rs996039356 Chr.4: 74038217-74038217 on Build GRCh38
rs748888221 Chr.4: 74038298-74038298 on Build GRCh38
rs781635868 Chr.4: 74038282-74038282 on Build GRCh38
rs760736689 Chr.4: 74038322-74038322 on Build GRCh38
rs1022034324 Chr.4: 74038218-74038218 on Build GRCh38
rs977756130 Chr.4: 74038233-74038233 on Build GRCh38
rs200035503 Chr.4: 74038324-74038324 on Build GRCh38
rs755983685 Chr.4: 74038170-74038170 on Build GRCh38
rs759175981 Chr.4: 74038136-74038136 on Build GRCh38
rs747052569 Chr.4: 74038279-74038279 on Build GRCh38
rs375380588 Chr.4: 74038285-74038285 on Build GRCh38
rs200373311 Chr.4: 74038259-74038259 on Build GRCh38
rs771736648 Chr.4: 74038318-74038318 on Build GRCh38
rs763480618 Chr.4: 74038240-74038240 on Build GRCh38
rs200228799 Chr.4: 74038306-74038306 on Build GRCh38
rs771173483 Chr.4: 74038129-74038129 on Build GRCh38
rs773263184 Chr.4: 74038320-74038320 on Build GRCh38
rs370332872 Chr.4: 74038197-74038197 on Build GRCh38
rs757258068 Chr.4: 74038157-74038157 on Build GRCh38
rs775129846 Chr.4: 74038150-74038150 on Build GRCh38
rs767207918 Chr.4: 74038158-74038158 on Build GRCh38
rs761638107 Chr.4: 74038249-74038249 on Build GRCh38
rs748406568 Chr.4: 74038192-74038192 on Build GRCh38
rs149422615 Chr.4: 74038151-74038151 on Build GRCh38
rs751429690 Chr.4: 74038155-74038155 on Build GRCh38
rs180805941 Chr.4: 74038317-74038317 on Build GRCh38
rs200604462 Chr.4: 74038323-74038323 on Build GRCh38
rs746167192 Chr.4: 74038212-74038212 on Build GRCh38
rs778809699 Chr.4: 74038219-74038219 on Build GRCh38
rs201940372 Chr.4: 74038135-74038135 on Build GRCh38
rs568286162 Chr.4: 74038252-74038252 on Build GRCh38
rs750297189 Chr.4: 74038169-74038169 on Build GRCh38
rs773975286 Chr.4: 74038247-74038247 on Build GRCh38
rs199744046 Chr.4: 74038319-74038319 on Build GRCh38
rs761297973 Chr.4: 74038126-74038126 on Build GRCh38
rs1050141386 Chr.4: 74038253-74038253 on Build GRCh38
rs773545719 Chr.4: 74038121-74038121 on Build GRCh38
rs201425298 Chr.4: 74038276-74038276 on Build GRCh38
rs764083321 Chr.4: 74038317-74038317 on Build GRCh38
rs768409602 Chr.4: 74038304-74038304 on Build GRCh38
rs202147554 Chr.4: 74038297-74038297 on Build GRCh38
rs773884964 Chr.4: 74038313-74038313 on Build GRCh38
rs755926685 Chr.4: 74038265-74038265 on Build GRCh38
rs777952677 Chr.4: 74038196-74038196 on Build GRCh38
rs771527719 Chr.4: 74038200-74038200 on Build GRCh38
rs764813946 Chr.4: 74038148-74038148 on Build GRCh38
rs770120324 Chr.4: 74038295-74038295 on Build GRCh38
rs758388814 Chr.4: 74038195-74038195 on Build GRCh38

View Full Product Details