Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49336868-49337133 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD37
Entrez Gene ID
951
Gene Name
CD37 molecule
Gene Aliases
GP52-40, TSPAN26
UniGene
Hs.166556
Gene Chromosome Location
Chr.19: 49335314-49340608 on Build GRCh38
Transcripts
NM_001040031.1 NM_001774.2 XM_005259435.3 XM_005259436.3 XM_011527542.2 XM_011527543.2 XM_011527544.2 XM_017027513.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3960272 c.220C>A p.L74M Chr.19: 49336986-49336986 on Build GRCh38
2752382 c.214G>A p.A72T Chr.19: 49336980-49336980 on Build GRCh38
3797373 c.213C>T p.I71I Chr.19: 49336979-49336979 on Build GRCh38
2752383 c.214G>A p.A72T Chr.19: 49336980-49336980 on Build GRCh38
3960271 c.220C>A p.L74M Chr.19: 49336986-49336986 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs949664201 Chr.19: 49337110-49337110 on Build GRCh38
rs376213637 Chr.19: 49337104-49337104 on Build GRCh38
rs751596329 Chr.19: 49336927-49336927 on Build GRCh38
rs777971755 Chr.19: 49337058-49337058 on Build GRCh38
rs995967646 Chr.19: 49336913-49336913 on Build GRCh38
rs755483220 Chr.19: 49337041-49337041 on Build GRCh38
rs140821686 Chr.19: 49336988-49336988 on Build GRCh38
rs747542770 Chr.19: 49336983-49336983 on Build GRCh38
rs764890793 Chr.19: 49336911-49336911 on Build GRCh38
rs767441803 Chr.19: 49336944-49336944 on Build GRCh38
rs374947160 Chr.19: 49336997-49336997 on Build GRCh38
rs780126495 Chr.19: 49337030-49337030 on Build GRCh38
rs961520984 Chr.19: 49336980-49336980 on Build GRCh38
rs759183644 Chr.19: 49337074-49337074 on Build GRCh38
rs149567989 Chr.19: 49336964-49336964 on Build GRCh38
rs753523959 Chr.19: 49337113-49337113 on Build GRCh38
rs771694041 Chr.19: 49337077-49337077 on Build GRCh38
rs201928399 Chr.19: 49337066-49337066 on Build GRCh38
rs150820452 Chr.19: 49336915-49336915 on Build GRCh38
rs761398616 Chr.19: 49336909-49336909 on Build GRCh38
rs201859925 Chr.19: 49336919-49336919 on Build GRCh38
rs769106160 Chr.19: 49336985-49336985 on Build GRCh38
rs369310469 Chr.19: 49337068-49337068 on Build GRCh38
rs780073603 Chr.19: 49336979-49336979 on Build GRCh38
rs147261609 Chr.19: 49336987-49336987 on Build GRCh38
rs755994691 Chr.19: 49337016-49337016 on Build GRCh38
rs375511347 Chr.19: 49337017-49337017 on Build GRCh38
rs751584302 Chr.19: 49337040-49337040 on Build GRCh38
rs752482696 Chr.19: 49336949-49336949 on Build GRCh38
rs766026576 Chr.19: 49336999-49336999 on Build GRCh38
rs200087875 Chr.19: 49337054-49337054 on Build GRCh38
rs1046096719 Chr.19: 49337111-49337111 on Build GRCh38
rs954775089 Chr.19: 49336954-49336954 on Build GRCh38
rs775543688 Chr.19: 49337080-49337080 on Build GRCh38
rs138165790 Chr.19: 49336950-49336950 on Build GRCh38
rs373001985 Chr.19: 49337101-49337101 on Build GRCh38
rs368729573 Chr.19: 49337006-49337006 on Build GRCh38
rs754934823 Chr.19: 49336936-49336936 on Build GRCh38
rs767661161 Chr.19: 49336995-49336995 on Build GRCh38
rs758551181 Chr.19: 49336967-49336967 on Build GRCh38
rs779259896 Chr.19: 49336955-49336955 on Build GRCh38
rs567113642 Chr.19: 49337003-49337003 on Build GRCh38
rs372181985 Chr.19: 49337009-49337009 on Build GRCh38
rs765795817 Chr.19: 49336920-49336920 on Build GRCh38
rs766085900 Chr.19: 49336898-49336898 on Build GRCh38
rs201004743 Chr.19: 49337055-49337055 on Build GRCh38
rs781752310 Chr.19: 49337045-49337045 on Build GRCh38
rs369527115 Chr.19: 49337026-49337026 on Build GRCh38

View Full Product Details