Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 9409733-9409982 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IPO7
Entrez Gene ID
10527
Gene Name
importin 7
Gene Aliases
Imp7, RANBP7
UniGene
Hs.744911
Gene Chromosome Location
Chr.11: 9384622-9448127 on Build GRCh38
Transcripts
NM_006391.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6427301 c.348C>A p.H116Q Chr.11: 9409955-9409955 on Build GRCh38
1662292 c.347A>C p.H116P Chr.11: 9409954-9409954 on Build GRCh38
4845434 c.321-1G>C p.? Chr.11: 9409927-9409927 on Build GRCh38
4431577 c.354C>G p.I118M Chr.11: 9409961-9409961 on Build GRCh38
4749740 c.343C>A p.H115N Chr.11: 9409950-9409950 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs956470410 Chr.11: 9409793-9409793 on Build GRCh38
rs769362885 Chr.11: 9409837-9409837 on Build GRCh38
rs1023932752 Chr.11: 9409806-9409806 on Build GRCh38
rs759383005 Chr.11: 9409905-9409905 on Build GRCh38
rs762736943 Chr.11: 9409897-9409897 on Build GRCh38
rs201024021 Chr.11: 9409932-9409932 on Build GRCh38
rs149757729 Chr.11: 9409804-9409804 on Build GRCh38
rs11042340 Chr.11: 9409939-9409939 on Build GRCh38
rs1026715792 Chr.11: 9409892-9409892 on Build GRCh38
rs151195385 Chr.11: 9409956-9409956 on Build GRCh38
rs750860600 Chr.11: 9409909-9409909 on Build GRCh38
rs571118982 Chr.11: 9409909-9409910 on Build GRCh38
rs746263888 Chr.11: 9409957-9409957 on Build GRCh38
rs979850666 Chr.11: 9409843-9409843 on Build GRCh38
rs559193323 Chr.11: 9409762-9409762 on Build GRCh38
rs776270222 Chr.11: 9409773-9409773 on Build GRCh38
rs140703485 Chr.11: 9409791-9409791 on Build GRCh38
rs1003644872 Chr.11: 9409775-9409775 on Build GRCh38
rs965970417 Chr.11: 9409854-9409854 on Build GRCh38
rs745627271 Chr.11: 9409835-9409835 on Build GRCh38
rs757664836 Chr.11: 9409929-9409929 on Build GRCh38
rs189601706 Chr.11: 9409763-9409763 on Build GRCh38
rs752965198 Chr.11: 9409914-9409914 on Build GRCh38
rs756387205 Chr.11: 9409910-9409910 on Build GRCh38
rs911826168 Chr.11: 9409850-9409850 on Build GRCh38
rs767894052 Chr.11: 9409908-9409908 on Build GRCh38
rs1034537442 Chr.11: 9409852-9409852 on Build GRCh38
rs756317955 Chr.11: 9409922-9409922 on Build GRCh38
rs144766309 Chr.11: 9409928-9409928 on Build GRCh38
rs919229790 Chr.11: 9409911-9409911 on Build GRCh38
rs891702008 Chr.11: 9409801-9409801 on Build GRCh38
rs766232113 Chr.11: 9409903-9409903 on Build GRCh38

View Full Product Details