Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 67656944-67657215 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYRK2
Entrez Gene ID
8445
Gene Name
dual specificity tyrosine phosphorylation regulated kinase 2
Gene Aliases
-
UniGene
Hs.173135
Gene Chromosome Location
Chr.12: 67648732-67665405 on Build GRCh38
Transcripts
NM_003583.3 NM_006482.2 XM_017020032.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4981322 c.222G>C p.M74I Chr.12: 67657129-67657129 on Build GRCh38
4044396 c.235C>T p.H79Y Chr.12: 67657142-67657142 on Build GRCh38
6328750 c.302G>A p.R101Q Chr.12: 67657209-67657209 on Build GRCh38
4147414 c.308T>A p.V103E Chr.12: 67657215-67657215 on Build GRCh38
1128485 c.207C>A p.G69G Chr.12: 67657114-67657114 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780452325 Chr.12: 67657127-67657127 on Build GRCh38
rs757234288 Chr.12: 67657089-67657089 on Build GRCh38
rs138020482 Chr.12: 67657175-67657175 on Build GRCh38
rs747348496 Chr.12: 67657132-67657132 on Build GRCh38
rs748856707 Chr.12: 67657144-67657144 on Build GRCh38
rs775477051 Chr.12: 67657157-67657157 on Build GRCh38
rs941860034 Chr.12: 67657026-67657026 on Build GRCh38
rs764431941 Chr.12: 67657176-67657176 on Build GRCh38
rs549647443 Chr.12: 67657052-67657052 on Build GRCh38
rs1015085446 Chr.12: 67656992-67656992 on Build GRCh38
rs772124910 Chr.12: 67657156-67657156 on Build GRCh38
rs745859869 Chr.12: 67657153-67657153 on Build GRCh38
rs961561536 Chr.12: 67657019-67657019 on Build GRCh38
rs530244321 Chr.12: 67657036-67657036 on Build GRCh38
rs986078966 Chr.12: 67657048-67657048 on Build GRCh38
rs756064741 Chr.12: 67657045-67657045 on Build GRCh38
rs878992942 Chr.12: 67657004-67657004 on Build GRCh38
rs879834687 Chr.12: 67656986-67656987 on Build GRCh38
rs1006003931 Chr.12: 67656982-67656982 on Build GRCh38
rs531415662 Chr.12: 67656995-67656995 on Build GRCh38
rs376543566 Chr.12: 67657056-67657056 on Build GRCh38
rs777366599 Chr.12: 67657140-67657140 on Build GRCh38
rs528905325 Chr.12: 67657147-67657147 on Build GRCh38
rs955628835 Chr.12: 67657033-67657033 on Build GRCh38
rs777721568 Chr.12: 67657067-67657067 on Build GRCh38
rs576473110 Chr.12: 67656971-67656971 on Build GRCh38
rs1029808577 Chr.12: 67657031-67657031 on Build GRCh38
rs546992961 Chr.12: 67657162-67657162 on Build GRCh38
rs199671497 Chr.12: 67657148-67657148 on Build GRCh38
rs994324956 Chr.12: 67657023-67657023 on Build GRCh38
rs758935204 Chr.12: 67657111-67657111 on Build GRCh38
rs968443245 Chr.12: 67657112-67657112 on Build GRCh38
rs750825894 Chr.12: 67657097-67657097 on Build GRCh38
rs764378967 Chr.12: 67657061-67657061 on Build GRCh38
rs924007523 Chr.12: 67657171-67657171 on Build GRCh38
rs761954038 Chr.12: 67657188-67657188 on Build GRCh38
rs200418247 Chr.12: 67657131-67657131 on Build GRCh38
rs376836219 Chr.12: 67657163-67657163 on Build GRCh38
rs909089798 Chr.12: 67657024-67657024 on Build GRCh38
rs765786973 Chr.12: 67657091-67657091 on Build GRCh38
rs749177348 Chr.12: 67657079-67657084 on Build GRCh38
rs567711118 Chr.12: 67657054-67657054 on Build GRCh38
rs900322867 Chr.12: 67657039-67657039 on Build GRCh38
rs755807656 Chr.12: 67657133-67657133 on Build GRCh38
rs780963887 Chr.12: 67657180-67657180 on Build GRCh38
rs753981412 Chr.12: 67657063-67657063 on Build GRCh38

View Full Product Details