Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 59320978-59321239 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDN3
Entrez Gene ID
1908
Gene Name
endothelin 3
Gene Aliases
ET-3, ET3, HSCR4, PPET3, WS4B
UniGene
Hs.1408
Gene Chromosome Location
Chr.20: 59300415-59325992 on Build GRCh38
Transcripts
NM_001302455.1 NM_001302456.1 NM_207032.2 NM_207033.2 NM_207034.2 XM_005260312.4 XM_005260313.4 XM_006723734.3 XM_011528655.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3668479 c.425C>T p.A142V Chr.20: 59321076-59321076 on Build GRCh38
5417446 c.534C>T p.D178D Chr.20: 59321185-59321185 on Build GRCh38
6651649 c.409C>T p.R137W Chr.20: 59321060-59321060 on Build GRCh38
3548375 c.401G>A p.G134E Chr.20: 59321052-59321052 on Build GRCh38
6651658 c.478G>A p.A160T Chr.20: 59321129-59321129 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs958504640 Chr.20: 59321026-59321026 on Build GRCh38
rs143232619 Chr.20: 59321152-59321152 on Build GRCh38
rs754800058 Chr.20: 59321009-59321009 on Build GRCh38
rs898401065 Chr.20: 59321074-59321074 on Build GRCh38
rs779957841 Chr.20: 59321161-59321161 on Build GRCh38
rs575758752 Chr.20: 59321146-59321146 on Build GRCh38
rs755789488 Chr.20: 59321042-59321042 on Build GRCh38
rs745314073 Chr.20: 59321131-59321131 on Build GRCh38
rs755843195 Chr.20: 59321117-59321117 on Build GRCh38
rs369295391 Chr.20: 59321121-59321121 on Build GRCh38
rs74315385 Chr.20: 59321158-59321158 on Build GRCh38
rs767453373 Chr.20: 59321186-59321186 on Build GRCh38
rs1050787473 Chr.20: 59321022-59321022 on Build GRCh38
rs762043350 Chr.20: 59321102-59321102 on Build GRCh38
rs145239588 Chr.20: 59321112-59321112 on Build GRCh38
rs765355566 Chr.20: 59321106-59321106 on Build GRCh38
rs74315384 Chr.20: 59321127-59321127 on Build GRCh38
rs374873073 Chr.20: 59321055-59321055 on Build GRCh38
rs752400458 Chr.20: 59321031-59321031 on Build GRCh38
rs781011983 Chr.20: 59321023-59321023 on Build GRCh38
rs779352707 Chr.20: 59321061-59321061 on Build GRCh38
rs774197901 Chr.20: 59321109-59321109 on Build GRCh38
rs756755957 Chr.20: 59321125-59321125 on Build GRCh38
rs151250216 Chr.20: 59321159-59321159 on Build GRCh38
rs778461734 Chr.20: 59321129-59321129 on Build GRCh38
rs775388121 Chr.20: 59321203-59321203 on Build GRCh38
rs767312223 Chr.20: 59321111-59321111 on Build GRCh38
rs374020215 Chr.20: 59321185-59321185 on Build GRCh38
rs777336048 Chr.20: 59321044-59321044 on Build GRCh38
rs867868088 Chr.20: 59321183-59321183 on Build GRCh38
rs576523143 Chr.20: 59321193-59321193 on Build GRCh38
rs149177260 Chr.20: 59321128-59321128 on Build GRCh38
rs536097197 Chr.20: 59321076-59321076 on Build GRCh38
rs377641145 Chr.20: 59321205-59321205 on Build GRCh38
rs769005371 Chr.20: 59321094-59321094 on Build GRCh38
rs34780366 Chr.20: 59321176-59321176 on Build GRCh38
rs748374290 Chr.20: 59321153-59321153 on Build GRCh38
rs187049336 Chr.20: 59321077-59321077 on Build GRCh38
rs758974269 Chr.20: 59321135-59321135 on Build GRCh38
rs372385465 Chr.20: 59321137-59321137 on Build GRCh38
rs776936096 Chr.20: 59321096-59321096 on Build GRCh38
rs34604265 Chr.20: 59321024-59321024 on Build GRCh38
rs753482233 Chr.20: 59321123-59321123 on Build GRCh38
rs772369425 Chr.20: 59321065-59321065 on Build GRCh38

View Full Product Details