Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 39382894-39383127 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAB2
Entrez Gene ID
1601
Gene Name
DAB2, clathrin adaptor protein
Gene Aliases
DOC-2, DOC2
UniGene
Hs.696631
Gene Chromosome Location
Chr.5: 39371674-39425233 on Build GRCh38
Transcripts
NM_001244871.1 NM_001343.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4983330 c.852C>G p.L284L Chr.5: 39383107-39383107 on Build GRCh38
5061093 c.919A>G p.T307A Chr.5: 39383040-39383040 on Build GRCh38
6190587 c.918G>A p.S306S Chr.5: 39383041-39383041 on Build GRCh38
187556 c.981G>A p.S327S Chr.5: 39382978-39382978 on Build GRCh38
737767 c.995G>T p.S332I Chr.5: 39382964-39382964 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754126227 Chr.5: 39383016-39383016 on Build GRCh38
rs748668338 Chr.5: 39382966-39382966 on Build GRCh38
rs751551355 Chr.5: 39382929-39382929 on Build GRCh38
rs760603265 Chr.5: 39383095-39383095 on Build GRCh38
rs200633845 Chr.5: 39383029-39383029 on Build GRCh38
rs1054018308 Chr.5: 39383021-39383021 on Build GRCh38
rs761765602 Chr.5: 39383007-39383007 on Build GRCh38
rs145888939 Chr.5: 39382956-39382956 on Build GRCh38
rs762576660 Chr.5: 39382988-39382988 on Build GRCh38
rs199558080 Chr.5: 39382969-39382969 on Build GRCh38
rs371072259 Chr.5: 39383037-39383037 on Build GRCh38
rs767671016 Chr.5: 39382935-39382935 on Build GRCh38
rs940338309 Chr.5: 39382967-39382967 on Build GRCh38
rs1033034084 Chr.5: 39383065-39383065 on Build GRCh38
rs766374522 Chr.5: 39383098-39383098 on Build GRCh38
rs774096033 Chr.5: 39383006-39383006 on Build GRCh38
rs756199635 Chr.5: 39382950-39382950 on Build GRCh38
rs866778073 Chr.5: 39383030-39383030 on Build GRCh38
rs927195939 Chr.5: 39383092-39383092 on Build GRCh38
rs773083361 Chr.5: 39383093-39383093 on Build GRCh38
rs34167601 Chr.5: 39383071-39383071 on Build GRCh38
rs750424788 Chr.5: 39382946-39382946 on Build GRCh38
rs779478292 Chr.5: 39383064-39383064 on Build GRCh38
rs755185803 Chr.5: 39383019-39383019 on Build GRCh38
rs200561912 Chr.5: 39382962-39382962 on Build GRCh38
rs550936413 Chr.5: 39382957-39382957 on Build GRCh38
rs1046605300 Chr.5: 39383051-39383051 on Build GRCh38
rs780180784 Chr.5: 39382951-39382951 on Build GRCh38
rs79344176 Chr.5: 39383073-39383073 on Build GRCh38
rs768449521 Chr.5: 39382992-39382992 on Build GRCh38
rs766470766 Chr.5: 39383011-39383011 on Build GRCh38
rs748973944 Chr.5: 39383067-39383067 on Build GRCh38
rs544112117 Chr.5: 39383072-39383072 on Build GRCh38
rs753430742 Chr.5: 39382978-39382978 on Build GRCh38
rs972589385 Chr.5: 39382960-39382960 on Build GRCh38
rs762772988 Chr.5: 39382919-39382919 on Build GRCh38
rs778220115 Chr.5: 39383030-39383032 on Build GRCh38
rs750346711 Chr.5: 39383008-39383008 on Build GRCh38
rs61748214 Chr.5: 39382971-39382971 on Build GRCh38
rs768253225 Chr.5: 39383068-39383068 on Build GRCh38
rs267600626 Chr.5: 39383042-39383042 on Build GRCh38
rs901530306 Chr.5: 39382970-39382970 on Build GRCh38
rs747544480 Chr.5: 39383049-39383049 on Build GRCh38
rs370444441 Chr.5: 39382964-39382964 on Build GRCh38
rs372564827 Chr.5: 39382921-39382921 on Build GRCh38
rs761675775 Chr.5: 39382930-39382930 on Build GRCh38
rs139705821 Chr.5: 39383058-39383058 on Build GRCh38
rs775069486 Chr.5: 39382987-39382987 on Build GRCh38

View Full Product Details