Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31811735-31812004 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPA1L
Entrez Gene ID
3305
Gene Name
heat shock protein family A (Hsp70) member 1 like
Gene Aliases
HSP70-1L, HSP70-HOM, HSP70T, hum70t
UniGene
Hs.690634
Gene Chromosome Location
Chr.6: 31809619-31815058 on Build GRCh38
Transcripts
NM_005527.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1294314 c.195C>G p.P65P Chr.6: 31811778-31811778 on Build GRCh38
1294315 c.186_187AA>GC p.M63L Chr.6: 31811786-31811787 on Build GRCh38
7096038 c.230G>A p.G77D Chr.6: 31811743-31811743 on Build GRCh38
1294312 c.219A>G p.K73K Chr.6: 31811754-31811754 on Build GRCh38
1294313 c.207T>G p.V69V Chr.6: 31811766-31811766 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758293750 Chr.6: 31811937-31811937 on Build GRCh38
rs775941765 Chr.6: 31811886-31811886 on Build GRCh38
rs751654845 Chr.6: 31811771-31811771 on Build GRCh38
rs777589589 Chr.6: 31811945-31811945 on Build GRCh38
rs748873814 Chr.6: 31811913-31811914 on Build GRCh38
rs772996658 Chr.6: 31811804-31811804 on Build GRCh38
rs541575734 Chr.6: 31811826-31811826 on Build GRCh38
rs754849874 Chr.6: 31811784-31811784 on Build GRCh38
rs762027845 Chr.6: 31811899-31811899 on Build GRCh38
rs746628579 Chr.6: 31811873-31811873 on Build GRCh38
rs151059204 Chr.6: 31811948-31811948 on Build GRCh38
rs373638266 Chr.6: 31811903-31811903 on Build GRCh38
rs189710538 Chr.6: 31811843-31811843 on Build GRCh38
rs770382651 Chr.6: 31811874-31811874 on Build GRCh38
rs772406755 Chr.6: 31811971-31811971 on Build GRCh38
rs778784868 Chr.6: 31811786-31811786 on Build GRCh38
rs760051391 Chr.6: 31811822-31811822 on Build GRCh38
rs757351933 Chr.6: 31811806-31811806 on Build GRCh38
rs763355013 Chr.6: 31811897-31811897 on Build GRCh38
rs749753182 Chr.6: 31811957-31811957 on Build GRCh38
rs141349016 Chr.6: 31811815-31811815 on Build GRCh38
rs765307007 Chr.6: 31811922-31811922 on Build GRCh38
rs9469057 Chr.6: 31811951-31811951 on Build GRCh38
rs757327606 Chr.6: 31811774-31811774 on Build GRCh38
rs369425515 Chr.6: 31811898-31811898 on Build GRCh38
rs780700552 Chr.6: 31811947-31811947 on Build GRCh38
rs150810052 Chr.6: 31811858-31811858 on Build GRCh38
rs768906825 Chr.6: 31811958-31811958 on Build GRCh38
rs777342418 Chr.6: 31811870-31811870 on Build GRCh38
rs764337395 Chr.6: 31811835-31811835 on Build GRCh38
rs760834707 Chr.6: 31811918-31811918 on Build GRCh38
rs752662698 Chr.6: 31811932-31811932 on Build GRCh38
rs779903526 Chr.6: 31811778-31811778 on Build GRCh38
rs373639301 Chr.6: 31811860-31811860 on Build GRCh38
rs767593530 Chr.6: 31811852-31811852 on Build GRCh38
rs747817123 Chr.6: 31811787-31811787 on Build GRCh38
rs1000603059 Chr.6: 31811825-31811825 on Build GRCh38
rs775886586 Chr.6: 31811832-31811832 on Build GRCh38
rs773538886 Chr.6: 31811974-31811974 on Build GRCh38
rs746543935 Chr.6: 31811805-31811805 on Build GRCh38
rs749200537 Chr.6: 31811782-31811782 on Build GRCh38
rs764284547 Chr.6: 31811767-31811767 on Build GRCh38
rs771910474 Chr.6: 31811803-31811803 on Build GRCh38
rs768310739 Chr.6: 31811948-31811949 on Build GRCh38
rs750574754 Chr.6: 31811916-31811916 on Build GRCh38
rs772862216 Chr.6: 31811796-31811796 on Build GRCh38
rs760889706 Chr.6: 31811975-31811975 on Build GRCh38
rs757305369 Chr.6: 31811845-31811845 on Build GRCh38
rs762535261 Chr.6: 31811840-31811840 on Build GRCh38
rs781443695 Chr.6: 31811762-31811763 on Build GRCh38
rs140839023 Chr.6: 31811966-31811966 on Build GRCh38
rs770349545 Chr.6: 31811880-31811880 on Build GRCh38
rs753725330 Chr.6: 31811854-31811854 on Build GRCh38
rs1007813910 Chr.6: 31811920-31811920 on Build GRCh38
rs147186823 Chr.6: 31811962-31811962 on Build GRCh38
rs372378700 Chr.6: 31811963-31811963 on Build GRCh38
rs903617843 Chr.6: 31811821-31811821 on Build GRCh38
rs763297872 Chr.6: 31811833-31811833 on Build GRCh38
rs892153963 Chr.6: 31811844-31811844 on Build GRCh38
rs767508406 Chr.6: 31811901-31811901 on Build GRCh38
rs745365008 Chr.6: 31811879-31811879 on Build GRCh38
rs201814236 Chr.6: 31811875-31811875 on Build GRCh38
rs530017660 Chr.6: 31811861-31811861 on Build GRCh38

View Full Product Details