Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 113118545-113118772 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F7
Entrez Gene ID
2155
Gene Name
coagulation factor VII
Gene Aliases
SPCA
UniGene
Hs.36989
Gene Chromosome Location
Chr.13: 113105773-113120681 on Build GRCh38
Transcripts
NM_000131.4 NM_001267554.1 NM_019616.3 NR_051961.1 XM_006719963.3 XM_011537474.2 XM_011537475.2 XM_011537476.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
945538 c.996G>A p.T332T Chr.13: 113118603-113118603 on Build GRCh38
7337635 c.859G>A p.E287K Chr.13: 113118673-113118673 on Build GRCh38
4619011 c.981G>A p.T327T Chr.13: 113118588-113118588 on Build GRCh38
6657783 c.1152G>A p.T384T Chr.13: 113118759-113118759 on Build GRCh38
3467360 c.1005C>T p.F335F Chr.13: 113118612-113118612 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756497287 Chr.13: 113118566-113118566 on Build GRCh38
rs140590804 Chr.13: 113118660-113118660 on Build GRCh38
rs149283257 Chr.13: 113118681-113118681 on Build GRCh38
rs121964927 Chr.13: 113118716-113118716 on Build GRCh38
rs574630461 Chr.13: 113118654-113118654 on Build GRCh38
rs779442604 Chr.13: 113118645-113118645 on Build GRCh38
rs377734791 Chr.13: 113118709-113118709 on Build GRCh38
rs201058276 Chr.13: 113118731-113118731 on Build GRCh38
rs904241671 Chr.13: 113118585-113118585 on Build GRCh38
rs754192993 Chr.13: 113118690-113118690 on Build GRCh38
rs121964926 Chr.13: 113118698-113118698 on Build GRCh38
rs749760143 Chr.13: 113118580-113118580 on Build GRCh38
rs200212201 Chr.13: 113118602-113118602 on Build GRCh38
rs139372641 Chr.13: 113118616-113118616 on Build GRCh38
rs748847358 Chr.13: 113118727-113118727 on Build GRCh38
rs769374785 Chr.13: 113118711-113118711 on Build GRCh38
rs374305125 Chr.13: 113118613-113118613 on Build GRCh38
rs778138366 Chr.13: 113118568-113118568 on Build GRCh38
rs768302905 Chr.13: 113118652-113118652 on Build GRCh38
rs137919286 Chr.13: 113118730-113118730 on Build GRCh38
rs747876824 Chr.13: 113118655-113118655 on Build GRCh38
rs963430078 Chr.13: 113118695-113118695 on Build GRCh38
rs576492410 Chr.13: 113118579-113118579 on Build GRCh38
rs754214942 Chr.13: 113118627-113118627 on Build GRCh38
rs769476482 Chr.13: 113118656-113118656 on Build GRCh38
rs534333896 Chr.13: 113118666-113118666 on Build GRCh38
rs945791616 Chr.13: 113118712-113118712 on Build GRCh38
rs779369836 Chr.13: 113118696-113118696 on Build GRCh38
rs768462253 Chr.13: 113118588-113118588 on Build GRCh38
rs752934007 Chr.13: 113118621-113118621 on Build GRCh38
rs36209567 Chr.13: 113118668-113118668 on Build GRCh38
rs761507772 Chr.13: 113118597-113118597 on Build GRCh38
rs868287262 Chr.13: 113118732-113118732 on Build GRCh38
rs387906508 Chr.13: 113118590-113118590 on Build GRCh38
rs762858015 Chr.13: 113118610-113118610 on Build GRCh38
rs774077974 Chr.13: 113118583-113118583 on Build GRCh38
rs928183869 Chr.13: 113118679-113118679 on Build GRCh38
rs537408773 Chr.13: 113118594-113118594 on Build GRCh38
rs895671158 Chr.13: 113118618-113118618 on Build GRCh38
rs757669648 Chr.13: 113118691-113118691 on Build GRCh38
rs778062451 Chr.13: 113118626-113118626 on Build GRCh38
rs202240468 Chr.13: 113118603-113118603 on Build GRCh38
rs759415146 Chr.13: 113118665-113118665 on Build GRCh38
rs750980786 Chr.13: 113118697-113118697 on Build GRCh38
rs770595177 Chr.13: 113118663-113118663 on Build GRCh38
rs752766242 Chr.13: 113118667-113118667 on Build GRCh38
rs757750790 Chr.13: 113118633-113118633 on Build GRCh38
rs960926421 Chr.13: 113118689-113118689 on Build GRCh38
rs746493506 Chr.13: 113118587-113118587 on Build GRCh38
rs146698837 Chr.13: 113118584-113118584 on Build GRCh38
rs747673406 Chr.13: 113118706-113118706 on Build GRCh38
rs756289549 Chr.13: 113118624-113118624 on Build GRCh38
rs746335937 Chr.13: 113118651-113118651 on Build GRCh38
rs368893306 Chr.13: 113118612-113118612 on Build GRCh38
rs774298365 Chr.13: 113118664-113118664 on Build GRCh38
rs142587242 Chr.13: 113118741-113118741 on Build GRCh38
rs3093267 Chr.13: 113118661-113118661 on Build GRCh38
rs767539514 Chr.13: 113118617-113118617 on Build GRCh38

View Full Product Details