Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 88448231-88448500 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXI3
Entrez Gene ID
344167
Gene Name
forkhead box I3
Gene Aliases
-
UniGene
Hs.716722
Gene Chromosome Location
Chr.2: 88448207-88452535 on Build GRCh38
Transcripts
NM_001135649.1

SNP Detail

SNP ID Chromosome Location
rs747609715 Chr.2: 88448312-88448312 on Build GRCh38
rs781657540 Chr.2: 88448342-88448342 on Build GRCh38
rs868321461 Chr.2: 88448439-88448439 on Build GRCh38
rs1052680274 Chr.2: 88448349-88448349 on Build GRCh38
rs571863821 Chr.2: 88448405-88448405 on Build GRCh38
rs201662120 Chr.2: 88448453-88448453 on Build GRCh38
rs770492023 Chr.2: 88448420-88448420 on Build GRCh38
rs547355868 Chr.2: 88448350-88448350 on Build GRCh38
rs769469825 Chr.2: 88448366-88448366 on Build GRCh38
rs761845956 Chr.2: 88448429-88448429 on Build GRCh38
rs925186048 Chr.2: 88448325-88448325 on Build GRCh38
rs898050540 Chr.2: 88448436-88448436 on Build GRCh38
rs905240619 Chr.2: 88448416-88448416 on Build GRCh38
rs539273096 Chr.2: 88448417-88448417 on Build GRCh38
rs568785675 Chr.2: 88448291-88448291 on Build GRCh38
rs752854592 Chr.2: 88448294-88448294 on Build GRCh38
rs370207846 Chr.2: 88448419-88448419 on Build GRCh38
rs767725521 Chr.2: 88448336-88448336 on Build GRCh38
rs752858246 Chr.2: 88448378-88448378 on Build GRCh38
rs965851622 Chr.2: 88448406-88448406 on Build GRCh38
rs963742773 Chr.2: 88448468-88448468 on Build GRCh38
rs528789316 Chr.2: 88448341-88448341 on Build GRCh38
rs768216591 Chr.2: 88448415-88448415 on Build GRCh38
rs200890046 Chr.2: 88448444-88448444 on Build GRCh38
rs1045046574 Chr.2: 88448408-88448408 on Build GRCh38
rs745918234 Chr.2: 88448365-88448365 on Build GRCh38
rs773079534 Chr.2: 88448451-88448451 on Build GRCh38
rs933758886 Chr.2: 88448476-88448476 on Build GRCh38
rs767872970 Chr.2: 88448342-88448353 on Build GRCh38
rs979385191 Chr.2: 88448304-88448304 on Build GRCh38
rs372022603 Chr.2: 88448382-88448382 on Build GRCh38
rs765558836 Chr.2: 88448460-88448460 on Build GRCh38
rs774808218 Chr.2: 88448297-88448297 on Build GRCh38
rs547264300 Chr.2: 88448259-88448259 on Build GRCh38
rs369600524 Chr.2: 88448457-88448457 on Build GRCh38
rs922394321 Chr.2: 88448458-88448458 on Build GRCh38
rs757531790 Chr.2: 88448321-88448321 on Build GRCh38
rs760031328 Chr.2: 88448303-88448303 on Build GRCh38
rs1034363987 Chr.2: 88448357-88448357 on Build GRCh38

View Full Product Details