Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 30679427-30679692 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSR
Entrez Gene ID
2936
Gene Name
glutathione-disulfide reductase
Gene Aliases
HEL-75, HEL-S-122m
UniGene
Hs.271510
Gene Chromosome Location
Chr.8: 30678061-30727969 on Build GRCh38
Transcripts
NM_000637.3 NM_001195102.1 NM_001195103.1 NM_001195104.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1623918 c.1555G>T p.V519F Chr.8: 30679534-30679534 on Build GRCh38
40824 c.1464G>A p.L488L Chr.8: 30679625-30679625 on Build GRCh38
6180633 c.1477G>C p.V493L Chr.8: 30679612-30679612 on Build GRCh38
5746858 c.1565G>A p.R522H Chr.8: 30679524-30679524 on Build GRCh38
2958213 c.1498A>C p.T500P Chr.8: 30679591-30679591 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746333168 Chr.8: 30679506-30679506 on Build GRCh38
rs200685394 Chr.8: 30679590-30679590 on Build GRCh38
rs775204777 Chr.8: 30679600-30679600 on Build GRCh38
rs757671437 Chr.8: 30679651-30679651 on Build GRCh38
rs989815814 Chr.8: 30679507-30679507 on Build GRCh38
rs369230085 Chr.8: 30679480-30679480 on Build GRCh38
rs143389671 Chr.8: 30679646-30679646 on Build GRCh38
rs759485069 Chr.8: 30679592-30679592 on Build GRCh38
rs147557976 Chr.8: 30679565-30679565 on Build GRCh38
rs373481457 Chr.8: 30679498-30679498 on Build GRCh38
rs896909854 Chr.8: 30679552-30679552 on Build GRCh38
rs763855107 Chr.8: 30679642-30679642 on Build GRCh38
rs377738754 Chr.8: 30679596-30679596 on Build GRCh38
rs149225584 Chr.8: 30679569-30679569 on Build GRCh38
rs765528479 Chr.8: 30679652-30679652 on Build GRCh38
rs779495845 Chr.8: 30679505-30679505 on Build GRCh38
rs1043070487 Chr.8: 30679463-30679463 on Build GRCh38
rs752407262 Chr.8: 30679496-30679496 on Build GRCh38
rs758486247 Chr.8: 30679524-30679524 on Build GRCh38
rs371380535 Chr.8: 30679603-30679603 on Build GRCh38
rs774242890 Chr.8: 30679588-30679588 on Build GRCh38
rs750774001 Chr.8: 30679660-30679660 on Build GRCh38
rs558512007 Chr.8: 30679550-30679550 on Build GRCh38
rs374914209 Chr.8: 30679572-30679572 on Build GRCh38
rs767063881 Chr.8: 30679493-30679493 on Build GRCh38
rs200171779 Chr.8: 30679481-30679481 on Build GRCh38
rs376582793 Chr.8: 30679473-30679473 on Build GRCh38
rs763647057 Chr.8: 30679499-30679499 on Build GRCh38
rs368833049 Chr.8: 30679589-30679589 on Build GRCh38
rs780167131 Chr.8: 30679534-30679534 on Build GRCh38
rs762626909 Chr.8: 30679470-30679470 on Build GRCh38
rs915133836 Chr.8: 30679519-30679519 on Build GRCh38
rs560445388 Chr.8: 30679483-30679483 on Build GRCh38
rs370972792 Chr.8: 30679621-30679621 on Build GRCh38
rs749133803 Chr.8: 30679586-30679586 on Build GRCh38
rs766391628 Chr.8: 30679474-30679474 on Build GRCh38
rs760070651 Chr.8: 30679630-30679630 on Build GRCh38
rs757538187 Chr.8: 30679500-30679500 on Build GRCh38
rs35768151 Chr.8: 30679475-30679475 on Build GRCh38

View Full Product Details