Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 33011636-33011869 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RHPN2
Entrez Gene ID
85415
Gene Name
rhophilin Rho GTPase binding protein 2
Gene Aliases
P76RBE, RHOBP
UniGene
Hs.466435
Gene Chromosome Location
Chr.19: 32978592-33064918 on Build GRCh38
Transcripts
NM_033103.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4841639 c.555C>T p.F185F Chr.19: 33011717-33011717 on Build GRCh38
5544186 c.529C>T p.Q177* Chr.19: 33011743-33011743 on Build GRCh38
6384186 c.476G>T p.R159L Chr.19: 33011796-33011796 on Build GRCh38
5019814 c.469-5T>C p.? Chr.19: 33011808-33011808 on Build GRCh38
2751311 c.488G>A p.R163Q Chr.19: 33011784-33011784 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs974975832 Chr.19: 33011691-33011691 on Build GRCh38
rs367645736 Chr.19: 33011783-33011783 on Build GRCh38
rs372301058 Chr.19: 33011797-33011797 on Build GRCh38
rs779530907 Chr.19: 33011728-33011728 on Build GRCh38
rs755180833 Chr.19: 33011675-33011675 on Build GRCh38
rs377498822 Chr.19: 33011654-33011654 on Build GRCh38
rs774310628 Chr.19: 33011773-33011773 on Build GRCh38
rs373249916 Chr.19: 33011793-33011793 on Build GRCh38
rs759412815 Chr.19: 33011661-33011662 on Build GRCh38
rs368366746 Chr.19: 33011796-33011796 on Build GRCh38
rs779804803 Chr.19: 33011807-33011807 on Build GRCh38
rs753111541 Chr.19: 33011697-33011697 on Build GRCh38
rs952304335 Chr.19: 33011705-33011705 on Build GRCh38
rs749127623 Chr.19: 33011729-33011729 on Build GRCh38
rs759547845 Chr.19: 33011785-33011785 on Build GRCh38
rs143511045 Chr.19: 33011792-33011792 on Build GRCh38
rs370371186 Chr.19: 33011703-33011703 on Build GRCh38
rs747881582 Chr.19: 33011847-33011847 on Build GRCh38
rs769487061 Chr.19: 33011723-33011723 on Build GRCh38
rs765182557 Chr.19: 33011842-33011845 on Build GRCh38
rs765319907 Chr.19: 33011695-33011695 on Build GRCh38
rs760589278 Chr.19: 33011721-33011721 on Build GRCh38
rs754124869 Chr.19: 33011659-33011659 on Build GRCh38
rs776705740 Chr.19: 33011784-33011784 on Build GRCh38
rs540549805 Chr.19: 33011715-33011715 on Build GRCh38
rs201914064 Chr.19: 33011811-33011811 on Build GRCh38
rs768562792 Chr.19: 33011828-33011828 on Build GRCh38
rs781734711 Chr.19: 33011799-33011799 on Build GRCh38
rs752092520 Chr.19: 33011704-33011704 on Build GRCh38
rs745316274 Chr.19: 33011722-33011722 on Build GRCh38
rs2288854 Chr.19: 33011808-33011808 on Build GRCh38
rs778664908 Chr.19: 33011843-33011843 on Build GRCh38
rs199535377 Chr.19: 33011735-33011735 on Build GRCh38
rs766414861 Chr.19: 33011658-33011658 on Build GRCh38
rs749982365 Chr.19: 33011803-33011803 on Build GRCh38
rs528621155 Chr.19: 33011711-33011711 on Build GRCh38
rs966303893 Chr.19: 33011686-33011686 on Build GRCh38
rs761734827 Chr.19: 33011774-33011774 on Build GRCh38

View Full Product Details