Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 143493558-143493831 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC9A9
Entrez Gene ID
285195
Gene Name
solute carrier family 9 member A9
Gene Aliases
AUTS16, NHE9
UniGene
Hs.302257
Gene Chromosome Location
Chr.3: 143265222-143848531 on Build GRCh38
Transcripts
NM_173653.3 XM_011512703.2 XM_017006202.1 XM_017006203.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1632873 c.1210A>T p.I404F Chr.3: 143493758-143493758 on Build GRCh38
382302 c.1263A>T p.L421L Chr.3: 143493705-143493705 on Build GRCh38
3588355 c.1308G>A p.M436I Chr.3: 143493660-143493660 on Build GRCh38
3125528 c.1268G>A p.R423Q Chr.3: 143493700-143493700 on Build GRCh38
5455144 c.1267C>T p.R423* Chr.3: 143493701-143493701 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199858098 Chr.3: 143493640-143493640 on Build GRCh38
rs755338869 Chr.3: 143493764-143493764 on Build GRCh38
rs368762649 Chr.3: 143493646-143493646 on Build GRCh38
rs748801537 Chr.3: 143493673-143493673 on Build GRCh38
rs760448809 Chr.3: 143493725-143493725 on Build GRCh38
rs775263877 Chr.3: 143493717-143493717 on Build GRCh38
rs372402210 Chr.3: 143493789-143493789 on Build GRCh38
rs375135790 Chr.3: 143493675-143493675 on Build GRCh38
rs121912597 Chr.3: 143493701-143493701 on Build GRCh38
rs762123497 Chr.3: 143493740-143493740 on Build GRCh38
rs773874013 Chr.3: 143493688-143493688 on Build GRCh38
rs767059595 Chr.3: 143493757-143493757 on Build GRCh38
rs776753475 Chr.3: 143493735-143493735 on Build GRCh38
rs755786076 Chr.3: 143493664-143493664 on Build GRCh38
rs765355968 Chr.3: 143493742-143493742 on Build GRCh38
rs376916789 Chr.3: 143493643-143493643 on Build GRCh38
rs184963714 Chr.3: 143493639-143493639 on Build GRCh38
rs748845522 Chr.3: 143493788-143493788 on Build GRCh38
rs778331615 Chr.3: 143493794-143493794 on Build GRCh38
rs780506256 Chr.3: 143493645-143493645 on Build GRCh38
rs747337131 Chr.3: 143493653-143493653 on Build GRCh38
rs777348368 Chr.3: 143493669-143493669 on Build GRCh38
rs781643252 Chr.3: 143493778-143493778 on Build GRCh38
rs764350984 Chr.3: 143493726-143493726 on Build GRCh38
rs770308974 Chr.3: 143493680-143493680 on Build GRCh38
rs765804835 Chr.3: 143493638-143493638 on Build GRCh38
rs112109858 Chr.3: 143493736-143493736 on Build GRCh38
rs181699849 Chr.3: 143493798-143493798 on Build GRCh38
rs368254745 Chr.3: 143493700-143493700 on Build GRCh38
rs754303991 Chr.3: 143493620-143493621 on Build GRCh38
rs1035618120 Chr.3: 143493649-143493649 on Build GRCh38
rs764293983 Chr.3: 143493617-143493617 on Build GRCh38
rs757326877 Chr.3: 143493636-143493636 on Build GRCh38

View Full Product Details