Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 143935861-143936092 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR2F1
Entrez Gene ID
26211
Gene Name
olfactory receptor family 2 subfamily F member 1 (gene/pseudogene)
Gene Aliases
7M1-2, OLF3, OR14-60, OR2F3, OR2F3P, OR2F4, OR2F5, OR7-139, OR7-140
UniGene
Hs.553595
Gene Chromosome Location
Chr.7: 143907152-143988540 on Build GRCh38
Transcripts
XM_011516020.2 NM_001004685.1

Gene Symbol
OR2F2
Entrez Gene ID
135948
Gene Name
olfactory receptor family 2 subfamily F member 2
Gene Aliases
OR7-1
UniGene
Hs.553596
Gene Chromosome Location
Chr.7: 143935233-143936186 on Build GRCh38
Transcripts
XM_011516020.2 NM_001004685.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3950112 c.800C>T p.S267L Chr.7: 143936032-143936032 on Build GRCh38
3635351 c.757G>A p.G253S Chr.7: 143935989-143935989 on Build GRCh38
421560 c.822C>G p.I274M Chr.7: 143936054-143936054 on Build GRCh38
3635352 c.786C>T p.P262P Chr.7: 143936018-143936018 on Build GRCh38
7319981 c.787C>T p.H263Y Chr.7: 143936019-143936019 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs558765768 Chr.7: 143935937-143935937 on Build GRCh38
rs779171600 Chr.7: 143936004-143936004 on Build GRCh38
rs752163600 Chr.7: 143935942-143935942 on Build GRCh38
rs199756492 Chr.7: 143935930-143935930 on Build GRCh38
rs768231854 Chr.7: 143935926-143935929 on Build GRCh38
rs777823367 Chr.7: 143935913-143935913 on Build GRCh38
rs757273782 Chr.7: 143935988-143935988 on Build GRCh38
rs201490066 Chr.7: 143935969-143935969 on Build GRCh38
rs772388791 Chr.7: 143936009-143936009 on Build GRCh38
rs754666549 Chr.7: 143935902-143935902 on Build GRCh38
rs749408795 Chr.7: 143935935-143935935 on Build GRCh38
rs201475989 Chr.7: 143935951-143935951 on Build GRCh38
rs775166890 Chr.7: 143936071-143936071 on Build GRCh38
rs1007173107 Chr.7: 143935974-143935974 on Build GRCh38
rs1050836668 Chr.7: 143935964-143935964 on Build GRCh38
rs750966120 Chr.7: 143935886-143935886 on Build GRCh38
rs369311135 Chr.7: 143935970-143935970 on Build GRCh38
rs755050627 Chr.7: 143935996-143935996 on Build GRCh38
rs773575356 Chr.7: 143936016-143936016 on Build GRCh38
rs777392051 Chr.7: 143936017-143936017 on Build GRCh38
rs748344565 Chr.7: 143936005-143936005 on Build GRCh38
rs773663131 Chr.7: 143935965-143935965 on Build GRCh38
rs764704431 Chr.7: 143936065-143936065 on Build GRCh38
rs866839513 Chr.7: 143935997-143935997 on Build GRCh38
rs758196267 Chr.7: 143935911-143935911 on Build GRCh38
rs749379251 Chr.7: 143935995-143935995 on Build GRCh38
rs187955599 Chr.7: 143935962-143935962 on Build GRCh38
rs752334026 Chr.7: 143935980-143935980 on Build GRCh38
rs199769704 Chr.7: 143935904-143935904 on Build GRCh38
rs755644378 Chr.7: 143935989-143935989 on Build GRCh38
rs765812619 Chr.7: 143935967-143935967 on Build GRCh38
rs371724112 Chr.7: 143936039-143936039 on Build GRCh38
rs777190443 Chr.7: 143936050-143936050 on Build GRCh38
rs375012448 Chr.7: 143935895-143935895 on Build GRCh38
rs376242048 Chr.7: 143935933-143935933 on Build GRCh38
rs749149031 Chr.7: 143936056-143936056 on Build GRCh38
rs768906164 Chr.7: 143935941-143935941 on Build GRCh38
rs929690509 Chr.7: 143935958-143935958 on Build GRCh38
rs774501246 Chr.7: 143935943-143935943 on Build GRCh38
rs764010721 Chr.7: 143935985-143935985 on Build GRCh38
rs967354196 Chr.7: 143936063-143936063 on Build GRCh38
rs747461176 Chr.7: 143936035-143936035 on Build GRCh38
rs368389594 Chr.7: 143935907-143935907 on Build GRCh38
rs61746066 Chr.7: 143935890-143935890 on Build GRCh38
rs61740239 Chr.7: 143935891-143935891 on Build GRCh38
rs773672641 Chr.7: 143936015-143936015 on Build GRCh38
rs13235235 Chr.7: 143936064-143936064 on Build GRCh38
rs4286866 Chr.7: 143935920-143935920 on Build GRCh38
rs758162035 Chr.7: 143935981-143935981 on Build GRCh38

View Full Product Details