Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 118039104-118039329 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB11FIP2
Entrez Gene ID
22841
Gene Name
RAB11 family interacting protein 2
Gene Aliases
Rab11-FIP2, nRip11
UniGene
Hs.173656
Gene Chromosome Location
Chr.10: 118004916-118046921 on Build GRCh38
Transcripts
NM_014904.2 XM_005269629.3 XM_017015926.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4012017 c.933A>G p.L311L Chr.10: 118039304-118039304 on Build GRCh38
915270 c.947C>A p.A316E Chr.10: 118039290-118039290 on Build GRCh38
6761190 c.959G>A p.R320K Chr.10: 118039278-118039278 on Build GRCh38
6761188 c.1067A>G p.K356R Chr.10: 118039170-118039170 on Build GRCh38
6505071 c.1030G>A p.E344K Chr.10: 118039207-118039207 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs551349191 Chr.10: 118039195-118039195 on Build GRCh38
rs762887793 Chr.10: 118039139-118039139 on Build GRCh38
rs754207808 Chr.10: 118039188-118039188 on Build GRCh38
rs374483786 Chr.10: 118039210-118039210 on Build GRCh38
rs745739576 Chr.10: 118039249-118039249 on Build GRCh38
rs543678419 Chr.10: 118039182-118039182 on Build GRCh38
rs763967221 Chr.10: 118039158-118039158 on Build GRCh38
rs755575274 Chr.10: 118039190-118039190 on Build GRCh38
rs200046106 Chr.10: 118039231-118039231 on Build GRCh38
rs775281814 Chr.10: 118039235-118039235 on Build GRCh38
rs747204294 Chr.10: 118039229-118039229 on Build GRCh38
rs750030820 Chr.10: 118039175-118039175 on Build GRCh38
rs756728056 Chr.10: 118039163-118039163 on Build GRCh38
rs187932637 Chr.10: 118039250-118039250 on Build GRCh38
rs752665382 Chr.10: 118039186-118039190 on Build GRCh38
rs754295912 Chr.10: 118039299-118039299 on Build GRCh38
rs760267092 Chr.10: 118039277-118039277 on Build GRCh38
rs1021266581 Chr.10: 118039200-118039200 on Build GRCh38
rs966990725 Chr.10: 118039211-118039211 on Build GRCh38
rs200498795 Chr.10: 118039236-118039236 on Build GRCh38
rs767247970 Chr.10: 118039165-118039165 on Build GRCh38
rs766089853 Chr.10: 118039293-118039293 on Build GRCh38
rs772034375 Chr.10: 118039212-118039212 on Build GRCh38
rs147770106 Chr.10: 118039148-118039148 on Build GRCh38
rs759923482 Chr.10: 118039180-118039183 on Build GRCh38
rs74353167 Chr.10: 118039300-118039300 on Build GRCh38
rs909938482 Chr.10: 118039203-118039203 on Build GRCh38
rs768617615 Chr.10: 118039244-118039244 on Build GRCh38
rs765384082 Chr.10: 118039184-118039184 on Build GRCh38
rs144228431 Chr.10: 118039141-118039141 on Build GRCh38
rs778128208 Chr.10: 118039220-118039220 on Build GRCh38
rs754341663 Chr.10: 118039164-118039167 on Build GRCh38
rs774247430 Chr.10: 118039246-118039246 on Build GRCh38
rs756567870 Chr.10: 118039186-118039186 on Build GRCh38
rs780296007 Chr.10: 118039187-118039187 on Build GRCh38
rs35192242 Chr.10: 118039142-118039142 on Build GRCh38
rs751097725 Chr.10: 118039161-118039161 on Build GRCh38

View Full Product Details