Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 112689651-112689888 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MOV10
Entrez Gene ID
4343
Gene Name
Mov10 RISC complex RNA helicase
Gene Aliases
fSAP113, gb110
UniGene
Hs.514941
Gene Chromosome Location
Chr.1: 112674312-112702377 on Build GRCh38
Transcripts
NM_001130079.2 NM_001286072.1 NM_001321324.1 NM_020963.4 XM_005270869.4 XM_005270870.4 XM_017001318.1 XM_017001319.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1600559 c.605G>T p.C202F Chr.1: 112689867-112689867 on Build GRCh38
1626313 c.577+1G>A p.? Chr.1: 112689651-112689651 on Build GRCh38
321731 c.622G>T p.G208C Chr.1: 112689884-112689884 on Build GRCh38
360691 c.621G>T p.V207V Chr.1: 112689883-112689883 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs768468337 Chr.1: 112689858-112689858 on Build GRCh38
rs780297800 Chr.1: 112689793-112689793 on Build GRCh38
rs3748659 Chr.1: 112689774-112689774 on Build GRCh38
rs199744267 Chr.1: 112689849-112689849 on Build GRCh38
rs1040640221 Chr.1: 112689755-112689755 on Build GRCh38
rs1050373454 Chr.1: 112689728-112689728 on Build GRCh38
rs769482206 Chr.1: 112689813-112689813 on Build GRCh38
rs774989722 Chr.1: 112689817-112689817 on Build GRCh38
rs376015380 Chr.1: 112689803-112689803 on Build GRCh38
rs375303480 Chr.1: 112689795-112689795 on Build GRCh38
rs888917957 Chr.1: 112689740-112689740 on Build GRCh38
rs956363616 Chr.1: 112689781-112689781 on Build GRCh38
rs776150168 Chr.1: 112689811-112689811 on Build GRCh38
rs570481139 Chr.1: 112689828-112689828 on Build GRCh38
rs766957320 Chr.1: 112689863-112689863 on Build GRCh38
rs759012200 Chr.1: 112689812-112689812 on Build GRCh38
rs946577487 Chr.1: 112689712-112689712 on Build GRCh38
rs561812974 Chr.1: 112689799-112689799 on Build GRCh38
rs904715929 Chr.1: 112689731-112689731 on Build GRCh38
rs144264245 Chr.1: 112689806-112689806 on Build GRCh38
rs555499055 Chr.1: 112689833-112689833 on Build GRCh38
rs901248906 Chr.1: 112689765-112689765 on Build GRCh38
rs59859800 Chr.1: 112689685-112689686 on Build GRCh38
rs762303121 Chr.1: 112689827-112689827 on Build GRCh38
rs756626417 Chr.1: 112689684-112689684 on Build GRCh38
rs1000600476 Chr.1: 112689768-112689768 on Build GRCh38
rs1008713125 Chr.1: 112689747-112689748 on Build GRCh38
rs755282300 Chr.1: 112689800-112689800 on Build GRCh38
rs527386779 Chr.1: 112689804-112689804 on Build GRCh38

View Full Product Details