Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 219915705-219915978 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC30A10
Entrez Gene ID
55532
Gene Name
solute carrier family 30 member 10
Gene Aliases
HMDPC, HMNDYT1, ZNT10, ZNT8, ZRC1, ZnT-10
UniGene
Hs.284450
Gene Chromosome Location
Chr.1: 219910398-219959754 on Build GRCh38
Transcripts
NM_018713.2 NR_046437.1 XM_006711437.3 XM_017001684.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1685851 c.1121G>A p.G374E Chr.1: 219915786-219915786 on Build GRCh38
1668485 c.1099C>T p.R367* Chr.1: 219915808-219915808 on Build GRCh38
7390427 c.1165G>C p.E389Q Chr.1: 219915742-219915742 on Build GRCh38
1689870 c.1121G>T p.G374V Chr.1: 219915786-219915786 on Build GRCh38
1689871 c.1000G>A p.E334K Chr.1: 219915907-219915907 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs922315420 Chr.1: 219915750-219915750 on Build GRCh38
rs752464591 Chr.1: 219915779-219915779 on Build GRCh38
rs746735960 Chr.1: 219915899-219915899 on Build GRCh38
rs889610512 Chr.1: 219915919-219915919 on Build GRCh38
rs149301485 Chr.1: 219915942-219915942 on Build GRCh38
rs780471993 Chr.1: 219915829-219915829 on Build GRCh38
rs749768848 Chr.1: 219915847-219915847 on Build GRCh38
rs776475615 Chr.1: 219915903-219915903 on Build GRCh38
rs943332891 Chr.1: 219915754-219915754 on Build GRCh38
rs776230339 Chr.1: 219915755-219915755 on Build GRCh38
rs751200217 Chr.1: 219915944-219915944 on Build GRCh38
rs993430155 Chr.1: 219915837-219915837 on Build GRCh38
rs762014945 Chr.1: 219915739-219915739 on Build GRCh38
rs758066830 Chr.1: 219915784-219915784 on Build GRCh38
rs768757012 Chr.1: 219915729-219915729 on Build GRCh38
rs765468456 Chr.1: 219915852-219915852 on Build GRCh38
rs375288113 Chr.1: 219915776-219915776 on Build GRCh38
rs200960383 Chr.1: 219915788-219915788 on Build GRCh38
rs774592416 Chr.1: 219915733-219915733 on Build GRCh38
rs779515587 Chr.1: 219915868-219915869 on Build GRCh38
rs756664488 Chr.1: 219915803-219915803 on Build GRCh38
rs766759426 Chr.1: 219915952-219915952 on Build GRCh38
rs770740586 Chr.1: 219915901-219915901 on Build GRCh38
rs1040426993 Chr.1: 219915764-219915764 on Build GRCh38
rs759019425 Chr.1: 219915756-219915756 on Build GRCh38
rs772219560 Chr.1: 219915869-219915869 on Build GRCh38
rs281860291 Chr.1: 219915861-219915861 on Build GRCh38
rs759514230 Chr.1: 219915913-219915913 on Build GRCh38
rs1033368999 Chr.1: 219915859-219915859 on Build GRCh38
rs762725299 Chr.1: 219915937-219915937 on Build GRCh38
rs766190303 Chr.1: 219915748-219915748 on Build GRCh38
rs959141901 Chr.1: 219915880-219915880 on Build GRCh38
rs202111121 Chr.1: 219915789-219915789 on Build GRCh38
rs778946938 Chr.1: 219915856-219915856 on Build GRCh38
rs750711888 Chr.1: 219915792-219915792 on Build GRCh38
rs772677934 Chr.1: 219915875-219915876 on Build GRCh38
rs775074045 Chr.1: 219915925-219915925 on Build GRCh38
rs746341448 Chr.1: 219915871-219915872 on Build GRCh38
rs886046001 Chr.1: 219915839-219915839 on Build GRCh38
rs991775564 Chr.1: 219915955-219915955 on Build GRCh38
rs773025168 Chr.1: 219915898-219915898 on Build GRCh38
rs756472368 Chr.1: 219915951-219915951 on Build GRCh38
rs564303811 Chr.1: 219915743-219915743 on Build GRCh38
rs779997407 Chr.1: 219915877-219915878 on Build GRCh38
rs887629558 Chr.1: 219915848-219915848 on Build GRCh38
rs748387183 Chr.1: 219915863-219915863 on Build GRCh38
rs371719088 Chr.1: 219915841-219915841 on Build GRCh38

View Full Product Details