Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 36975547-36975820 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTCH1
Entrez Gene ID
23787
Gene Name
mitochondrial carrier 1
Gene Aliases
CGI-64, PIG60, PSAP, SLC25A49
UniGene
Hs.485262
Gene Chromosome Location
Chr.6: 36968135-36986551 on Build GRCh38
Transcripts
NM_001271641.1 NM_014341.2 NR_130739.1 XM_005248978.1 XM_005248979.1 XM_011514450.1 XM_017010642.1 XM_017010643.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1727710 c.751G>T p.G251* Chr.6: 36975668-36975668 on Build GRCh38
7333073 c.559G>T p.G187* Chr.6: 36975668-36975668 on Build GRCh38
1727711 c.751G>T p.G251* Chr.6: 36975668-36975668 on Build GRCh38
7333075 c.243G>T p.L81L Chr.6: 36975668-36975668 on Build GRCh38
7333074 c.283G>T p.G95* Chr.6: 36975668-36975668 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs148957702 Chr.6: 36975594-36975594 on Build GRCh38
rs771813210 Chr.6: 36975700-36975700 on Build GRCh38
rs192687584 Chr.6: 36975795-36975795 on Build GRCh38
rs939117476 Chr.6: 36975582-36975582 on Build GRCh38
rs183558462 Chr.6: 36975659-36975659 on Build GRCh38
rs375839256 Chr.6: 36975764-36975764 on Build GRCh38
rs369033339 Chr.6: 36975570-36975570 on Build GRCh38
rs188617109 Chr.6: 36975746-36975746 on Build GRCh38
rs749072289 Chr.6: 36975642-36975642 on Build GRCh38
rs779767732 Chr.6: 36975645-36975645 on Build GRCh38
rs917975632 Chr.6: 36975780-36975780 on Build GRCh38
rs1056170134 Chr.6: 36975592-36975592 on Build GRCh38
rs73422776 Chr.6: 36975778-36975778 on Build GRCh38
rs769993445 Chr.6: 36975745-36975745 on Build GRCh38
rs1013122189 Chr.6: 36975604-36975604 on Build GRCh38
rs768730375 Chr.6: 36975747-36975747 on Build GRCh38
rs945445543 Chr.6: 36975786-36975786 on Build GRCh38
rs115240884 Chr.6: 36975799-36975799 on Build GRCh38
rs778749264 Chr.6: 36975672-36975672 on Build GRCh38
rs748071779 Chr.6: 36975681-36975681 on Build GRCh38
rs888715967 Chr.6: 36975640-36975640 on Build GRCh38
rs17554163 Chr.6: 36975755-36975755 on Build GRCh38
rs961426930 Chr.6: 36975733-36975733 on Build GRCh38
rs772961439 Chr.6: 36975727-36975727 on Build GRCh38
rs750135968 Chr.6: 36975636-36975636 on Build GRCh38
rs977697815 Chr.6: 36975791-36975791 on Build GRCh38
rs372986933 Chr.6: 36975625-36975625 on Build GRCh38
rs369542410 Chr.6: 36975660-36975660 on Build GRCh38
rs368651137 Chr.6: 36975777-36975777 on Build GRCh38
rs2293386 Chr.6: 36975797-36975797 on Build GRCh38
rs537820523 Chr.6: 36975620-36975620 on Build GRCh38
rs752062926 Chr.6: 36975624-36975624 on Build GRCh38
rs777808773 Chr.6: 36975641-36975641 on Build GRCh38
rs1015670965 Chr.6: 36975671-36975671 on Build GRCh38
rs374210762 Chr.6: 36975634-36975634 on Build GRCh38
rs2293385 Chr.6: 36975737-36975737 on Build GRCh38
rs200342013 Chr.6: 36975637-36975637 on Build GRCh38

View Full Product Details