Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 212920-213163 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RIC8A
Entrez Gene ID
60626
Gene Name
RIC8 guanine nucleotide exchange factor A
Gene Aliases
RIC8
UniGene
Hs.592292
Gene Chromosome Location
Chr.11: 207511-215175 on Build GRCh38
Transcripts
NM_001286134.1 NM_021932.5 XM_005253052.3 XM_017018107.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4525886 c.1360G>A p.E454K Chr.11: 212968-212968 on Build GRCh38
6563199 c.1325_1326insAA p.E443fs*18 Chr.11: 212933-212934 on Build GRCh38
248718 c.1340C>A p.T447K Chr.11: 212948-212948 on Build GRCh38
6576061 c.1351G>A p.E451K Chr.11: 212959-212959 on Build GRCh38
1739771 c.1354T>A p.Y452N Chr.11: 212962-212962 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1056767988 Chr.11: 212943-212943 on Build GRCh38
rs748109810 Chr.11: 213013-213013 on Build GRCh38
rs888502244 Chr.11: 213015-213015 on Build GRCh38
rs1007763937 Chr.11: 213050-213050 on Build GRCh38
rs376357285 Chr.11: 212952-212952 on Build GRCh38
rs777845594 Chr.11: 212964-212964 on Build GRCh38
rs372169188 Chr.11: 213030-213030 on Build GRCh38
rs757982968 Chr.11: 212955-212955 on Build GRCh38
rs1019604374 Chr.11: 213078-213078 on Build GRCh38
rs779208651 Chr.11: 213012-213012 on Build GRCh38
rs200428378 Chr.11: 213031-213031 on Build GRCh38
rs966284894 Chr.11: 213091-213091 on Build GRCh38
rs781170894 Chr.11: 213004-213004 on Build GRCh38
rs1012565631 Chr.11: 213075-213075 on Build GRCh38
rs772207413 Chr.11: 213026-213026 on Build GRCh38
rs768710602 Chr.11: 213009-213009 on Build GRCh38
rs751602258 Chr.11: 212989-212989 on Build GRCh38
rs142035641 Chr.11: 213028-213028 on Build GRCh38
rs988356833 Chr.11: 213139-213139 on Build GRCh38
rs534387898 Chr.11: 212990-212990 on Build GRCh38
rs1003874120 Chr.11: 212953-212953 on Build GRCh38
rs556813572 Chr.11: 213132-213132 on Build GRCh38
rs553963905 Chr.11: 213005-213005 on Build GRCh38
rs1036666035 Chr.11: 212965-212965 on Build GRCh38
rs1024916086 Chr.11: 213123-213123 on Build GRCh38

View Full Product Details