Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 55773992-55774265 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR5D13
Entrez Gene ID
390142
Gene Name
olfactory receptor family 5 subfamily D member 13 (gene/pseudogene)
Gene Aliases
-
UniGene
Hs.553743
Gene Chromosome Location
Chr.11: 55773438-55774382 on Build GRCh38
Transcripts
NM_001001967.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5660439 c.788T>C p.V263A Chr.11: 55774225-55774225 on Build GRCh38
3722900 c.756C>G p.I252M Chr.11: 55774193-55774193 on Build GRCh38
1293377 c.588C>T p.I196I Chr.11: 55774025-55774025 on Build GRCh38
4406308 c.695C>T p.S232F Chr.11: 55774132-55774132 on Build GRCh38
6881630 c.743C>T p.T248I Chr.11: 55774180-55774180 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762561286 Chr.11: 55774078-55774078 on Build GRCh38
rs760258274 Chr.11: 55774085-55774085 on Build GRCh38
rs753203578 Chr.11: 55774128-55774128 on Build GRCh38
rs192984177 Chr.11: 55774142-55774142 on Build GRCh38
rs756249798 Chr.11: 55774059-55774059 on Build GRCh38
rs772981785 Chr.11: 55774169-55774169 on Build GRCh38
rs766729051 Chr.11: 55774116-55774116 on Build GRCh38
rs749525915 Chr.11: 55774065-55774065 on Build GRCh38
rs771883171 Chr.11: 55774074-55774074 on Build GRCh38
rs542711718 Chr.11: 55774234-55774234 on Build GRCh38
rs201933246 Chr.11: 55774045-55774045 on Build GRCh38
rs7124871 Chr.11: 55774144-55774144 on Build GRCh38
rs375224277 Chr.11: 55774053-55774053 on Build GRCh38
rs750662334 Chr.11: 55774222-55774222 on Build GRCh38
rs774012401 Chr.11: 55774120-55774120 on Build GRCh38
rs757388285 Chr.11: 55774134-55774134 on Build GRCh38
rs76632744 Chr.11: 55774168-55774168 on Build GRCh38
rs777852254 Chr.11: 55774129-55774129 on Build GRCh38
rs763398517 Chr.11: 55774183-55774184 on Build GRCh38
rs777900167 Chr.11: 55774060-55774060 on Build GRCh38
rs74548274 Chr.11: 55774029-55774029 on Build GRCh38
rs769315589 Chr.11: 55774161-55774161 on Build GRCh38
rs768237305 Chr.11: 55774147-55774147 on Build GRCh38
rs146084338 Chr.11: 55774033-55774033 on Build GRCh38
rs759749073 Chr.11: 55774180-55774180 on Build GRCh38
rs781746014 Chr.11: 55774052-55774052 on Build GRCh38
rs763695415 Chr.11: 55774091-55774091 on Build GRCh38
rs546719716 Chr.11: 55774018-55774018 on Build GRCh38
rs745713266 Chr.11: 55774071-55774071 on Build GRCh38
rs761429065 Chr.11: 55774160-55774160 on Build GRCh38
rs369729738 Chr.11: 55774143-55774143 on Build GRCh38
rs766670995 Chr.11: 55774039-55774039 on Build GRCh38
rs774044390 Chr.11: 55774070-55774070 on Build GRCh38
rs760000360 Chr.11: 55774124-55774124 on Build GRCh38
rs557354554 Chr.11: 55774119-55774119 on Build GRCh38
rs776369858 Chr.11: 55774095-55774095 on Build GRCh38
rs758210309 Chr.11: 55774233-55774233 on Build GRCh38
rs764623697 Chr.11: 55774190-55774190 on Build GRCh38
rs868211568 Chr.11: 55774163-55774163 on Build GRCh38
rs148672786 Chr.11: 55774126-55774126 on Build GRCh38
rs199573658 Chr.11: 55774024-55774024 on Build GRCh38
rs867423283 Chr.11: 55774082-55774082 on Build GRCh38
rs201249404 Chr.11: 55774138-55774138 on Build GRCh38
rs757417473 Chr.11: 55774209-55774209 on Build GRCh38
rs753152807 Chr.11: 55774186-55774186 on Build GRCh38
rs753838527 Chr.11: 55774204-55774204 on Build GRCh38
rs561869024 Chr.11: 55774159-55774159 on Build GRCh38
rs761398608 Chr.11: 55774114-55774114 on Build GRCh38
rs779000685 Chr.11: 55774220-55774220 on Build GRCh38
rs991150579 Chr.11: 55774208-55774208 on Build GRCh38
rs770710079 Chr.11: 55774066-55774066 on Build GRCh38
rs754018545 Chr.11: 55774132-55774132 on Build GRCh38
rs761201512 Chr.11: 55774188-55774188 on Build GRCh38
rs775106926 Chr.11: 55774026-55774026 on Build GRCh38
rs767879042 Chr.11: 55774182-55774182 on Build GRCh38

View Full Product Details