Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 29200423-29200684 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MECR
Entrez Gene ID
51102
Gene Name
mitochondrial trans-2-enoyl-CoA reductase
Gene Aliases
CGI-63, FASN2B, NRBF1
UniGene
Hs.183646
Gene Chromosome Location
Chr.1: 29192657-29230958 on Build GRCh38
Transcripts
NM_001024732.2 NM_016011.3 XM_005245885.2 XM_005245887.2 XM_011541539.1 XM_011541540.1 XM_011541541.1 XM_011541543.1 XM_011541545.1 XM_011541546.1 XM_011541547.1 XM_011541549.1 XM_011541550.1 XM_011541553.1 XM_011541554.1 XM_017001411.1 XM_017001412.1 XM_017001413.1 XM_017001414.1 XM_017001415.1 XM_017001416.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3746926 c.492C>T p.S164S Chr.1: 29200516-29200516 on Build GRCh38
6710561 c.821G>A p.R274Q Chr.1: 29200525-29200525 on Build GRCh38
6710562 c.483G>A p.A161A Chr.1: 29200525-29200525 on Build GRCh38
908048 c.830C>T p.A277V Chr.1: 29200516-29200516 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs748488801 Chr.1: 29200603-29200603 on Build GRCh38
rs535321004 Chr.1: 29200595-29200595 on Build GRCh38
rs771606009 Chr.1: 29200634-29200634 on Build GRCh38
rs768150493 Chr.1: 29200544-29200544 on Build GRCh38
rs529474105 Chr.1: 29200509-29200509 on Build GRCh38
rs758280997 Chr.1: 29200599-29200599 on Build GRCh38
rs769426672 Chr.1: 29200558-29200558 on Build GRCh38
rs148978800 Chr.1: 29200516-29200516 on Build GRCh38
rs752950079 Chr.1: 29200581-29200581 on Build GRCh38
rs367854119 Chr.1: 29200485-29200485 on Build GRCh38
rs536752565 Chr.1: 29200653-29200653 on Build GRCh38
rs756421370 Chr.1: 29200513-29200514 on Build GRCh38
rs138711412 Chr.1: 29200635-29200635 on Build GRCh38
rs776328593 Chr.1: 29200546-29200546 on Build GRCh38
rs1049957605 Chr.1: 29200555-29200555 on Build GRCh38
rs763497928 Chr.1: 29200563-29200563 on Build GRCh38
rs746692331 Chr.1: 29200526-29200526 on Build GRCh38
rs369359563 Chr.1: 29200502-29200502 on Build GRCh38
rs771857782 Chr.1: 29200520-29200520 on Build GRCh38
rs371988968 Chr.1: 29200612-29200612 on Build GRCh38
rs750956942 Chr.1: 29200476-29200476 on Build GRCh38
rs747674242 Chr.1: 29200547-29200547 on Build GRCh38
rs369875973 Chr.1: 29200627-29200627 on Build GRCh38
rs899070369 Chr.1: 29200451-29200451 on Build GRCh38
rs774899584 Chr.1: 29200639-29200639 on Build GRCh38
rs1037485489 Chr.1: 29200618-29200618 on Build GRCh38
rs2503016 Chr.1: 29200633-29200633 on Build GRCh38
rs764093603 Chr.1: 29200592-29200592 on Build GRCh38
rs747772734 Chr.1: 29200626-29200626 on Build GRCh38
rs145192716 Chr.1: 29200574-29200574 on Build GRCh38
rs779925640 Chr.1: 29200601-29200601 on Build GRCh38
rs762409659 Chr.1: 29200470-29200470 on Build GRCh38
rs1041340876 Chr.1: 29200504-29200504 on Build GRCh38
rs34835902 Chr.1: 29200573-29200573 on Build GRCh38
rs778879465 Chr.1: 29200515-29200515 on Build GRCh38
rs140895510 Chr.1: 29200525-29200525 on Build GRCh38
rs375317652 Chr.1: 29200613-29200613 on Build GRCh38
rs765898406 Chr.1: 29200475-29200475 on Build GRCh38
rs755107970 Chr.1: 29200494-29200494 on Build GRCh38
rs565992413 Chr.1: 29200464-29200464 on Build GRCh38
rs112436480 Chr.1: 29200498-29200498 on Build GRCh38
rs995827217 Chr.1: 29200458-29200458 on Build GRCh38
rs778141586 Chr.1: 29200551-29200552 on Build GRCh38
rs756251873 Chr.1: 29200585-29200585 on Build GRCh38
rs768883736 Chr.1: 29200552-29200552 on Build GRCh38
rs766875421 Chr.1: 29200565-29200565 on Build GRCh38
rs763481343 Chr.1: 29200482-29200482 on Build GRCh38

View Full Product Details