Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150150879-150151134 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLEKHO1
Entrez Gene ID
51177
Gene Name
pleckstrin homology domain containing O1
Gene Aliases
CKIP-1, CKIP1, JBP, OC120
UniGene
Hs.438824
Gene Chromosome Location
Chr.1: 150149434-150160065 on Build GRCh38
Transcripts
NM_001304722.1 NM_001304723.1 NM_001304724.1 NM_016274.5 XM_017001420.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
163555 c.32G>A p.G11E Chr.1: 150150913-150150913 on Build GRCh38
3711132 c.39G>T p.Q13H Chr.1: 150150920-150150920 on Build GRCh38
6261669 c.82G>T p.V28F Chr.1: 150150963-150150963 on Build GRCh38
284095 c.135T>C p.Y45Y Chr.1: 150151016-150151016 on Build GRCh38
3976099 c.76G>A p.G26S Chr.1: 150150957-150150957 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782121103 Chr.1: 150151067-150151067 on Build GRCh38
rs782712497 Chr.1: 150150959-150150959 on Build GRCh38
rs782360361 Chr.1: 150151068-150151068 on Build GRCh38
rs782641113 Chr.1: 150150902-150150902 on Build GRCh38
rs782289064 Chr.1: 150150915-150150915 on Build GRCh38
rs1045480594 Chr.1: 150151115-150151115 on Build GRCh38
rs782333775 Chr.1: 150151032-150151032 on Build GRCh38
rs782805712 Chr.1: 150151073-150151073 on Build GRCh38
rs782299637 Chr.1: 150151040-150151040 on Build GRCh38
rs587736199 Chr.1: 150151013-150151013 on Build GRCh38
rs782713489 Chr.1: 150151084-150151084 on Build GRCh38
rs199707212 Chr.1: 150151108-150151108 on Build GRCh38
rs782571742 Chr.1: 150151036-150151036 on Build GRCh38
rs781870851 Chr.1: 150151012-150151012 on Build GRCh38
rs782546778 Chr.1: 150150995-150150995 on Build GRCh38
rs781959039 Chr.1: 150151069-150151069 on Build GRCh38
rs919003315 Chr.1: 150150997-150150997 on Build GRCh38
rs782774322 Chr.1: 150151076-150151077 on Build GRCh38
rs781998715 Chr.1: 150150917-150150917 on Build GRCh38
rs781959956 Chr.1: 150150940-150150940 on Build GRCh38
rs371203990 Chr.1: 150151112-150151112 on Build GRCh38
rs948306977 Chr.1: 150151102-150151102 on Build GRCh38
rs951581847 Chr.1: 150151019-150151019 on Build GRCh38
rs782126834 Chr.1: 150151095-150151095 on Build GRCh38
rs782500984 Chr.1: 150150953-150150953 on Build GRCh38
rs376545625 Chr.1: 150150967-150150967 on Build GRCh38
rs74124329 Chr.1: 150150908-150150908 on Build GRCh38
rs781884859 Chr.1: 150151074-150151074 on Build GRCh38
rs782013254 Chr.1: 150151045-150151045 on Build GRCh38
rs147235795 Chr.1: 150150948-150150948 on Build GRCh38
rs782448763 Chr.1: 150150907-150150907 on Build GRCh38
rs910148586 Chr.1: 150151094-150151094 on Build GRCh38
rs782241044 Chr.1: 150150921-150150921 on Build GRCh38
rs782041653 Chr.1: 150151071-150151071 on Build GRCh38
rs782070423 Chr.1: 150150941-150150941 on Build GRCh38
rs782153941 Chr.1: 150150904-150150905 on Build GRCh38
rs782763685 Chr.1: 150151105-150151105 on Build GRCh38
rs587596528 Chr.1: 150151023-150151023 on Build GRCh38
rs144644257 Chr.1: 150151042-150151042 on Build GRCh38
rs782660189 Chr.1: 150151008-150151008 on Build GRCh38

View Full Product Details