Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 34409430-34409655 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDCD2L
Entrez Gene ID
84306
Gene Name
programmed cell death 2 like
Gene Aliases
-
UniGene
Hs.515344
Gene Chromosome Location
Chr.19: 34404398-34426168 on Build GRCh38
Transcripts
NM_032346.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
292433 c.659T>C p.I220T Chr.19: 34409483-34409483 on Build GRCh38
394178 c.658A>G p.I220V Chr.19: 34409482-34409482 on Build GRCh38
994763 c.652G>T p.E218* Chr.19: 34409476-34409476 on Build GRCh38
6896326 c.652G>C p.E218Q Chr.19: 34409476-34409476 on Build GRCh38
1318322 c.641A>G p.Y214C Chr.19: 34409465-34409465 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746523850 Chr.19: 34409508-34409508 on Build GRCh38
rs753764732 Chr.19: 34409533-34409536 on Build GRCh38
rs758183762 Chr.19: 34409483-34409483 on Build GRCh38
rs769597269 Chr.19: 34409527-34409527 on Build GRCh38
rs760821047 Chr.19: 34409551-34409551 on Build GRCh38
rs1021366187 Chr.19: 34409621-34409621 on Build GRCh38
rs778389275 Chr.19: 34409476-34409476 on Build GRCh38
rs750087893 Chr.19: 34409510-34409511 on Build GRCh38
rs755390769 Chr.19: 34409465-34409465 on Build GRCh38
rs762004866 Chr.19: 34409545-34409545 on Build GRCh38
rs2161473 Chr.19: 34409455-34409455 on Build GRCh38
rs770975776 Chr.19: 34409562-34409580 on Build GRCh38
rs370218371 Chr.19: 34409495-34409495 on Build GRCh38
rs778805460 Chr.19: 34409545-34409546 on Build GRCh38
rs760011470 Chr.19: 34409560-34409560 on Build GRCh38
rs373032906 Chr.19: 34409461-34409461 on Build GRCh38
rs747255056 Chr.19: 34409556-34409557 on Build GRCh38
rs775310315 Chr.19: 34409539-34409539 on Build GRCh38
rs760216585 Chr.19: 34409517-34409520 on Build GRCh38
rs772344250 Chr.19: 34409550-34409550 on Build GRCh38
rs758167729 Chr.19: 34409549-34409550 on Build GRCh38
rs747782438 Chr.19: 34409477-34409477 on Build GRCh38
rs766744887 Chr.19: 34409556-34409556 on Build GRCh38
rs796947345 Chr.19: 34409545-34409546 on Build GRCh38
rs531785553 Chr.19: 34409525-34409525 on Build GRCh38
rs778036256 Chr.19: 34409554-34409558 on Build GRCh38
rs754983563 Chr.19: 34409542-34409543 on Build GRCh38
rs911682530 Chr.19: 34409496-34409496 on Build GRCh38
rs745612586 Chr.19: 34409526-34409526 on Build GRCh38
rs770719960 Chr.19: 34409515-34409515 on Build GRCh38
rs548781456 Chr.19: 34409557-34409557 on Build GRCh38
rs765875393 Chr.19: 34409530-34409531 on Build GRCh38

View Full Product Details