Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 151100908-151101181 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPS3A
Entrez Gene ID
6189
Gene Name
ribosomal protein S3A
Gene Aliases
FTE1, MFTL, S3A
UniGene
Hs.356572
Gene Chromosome Location
Chr.4: 151099573-151104652 on Build GRCh38
Transcripts
NM_001006.4 NM_001267699.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4852538 c.232G>A p.E78K Chr.4: 151101040-151101040 on Build GRCh38
6288595 c.300C>T p.F100F Chr.4: 151101108-151101108 on Build GRCh38
1052041 c.252C>A p.F84L Chr.4: 151101060-151101060 on Build GRCh38
6278633 c.279T>C p.G93G Chr.4: 151101087-151101087 on Build GRCh38
5023968 c.297_298insA p.F100fs*9 Chr.4: 151101105-151101106 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs753943493 Chr.4: 151100970-151100970 on Build GRCh38
rs774176378 Chr.4: 151101108-151101108 on Build GRCh38
rs777505592 Chr.4: 151101086-151101086 on Build GRCh38
rs372732815 Chr.4: 151100955-151100955 on Build GRCh38
rs541190765 Chr.4: 151100998-151100998 on Build GRCh38
rs756074679 Chr.4: 151101078-151101078 on Build GRCh38
rs775982029 Chr.4: 151101132-151101132 on Build GRCh38
rs760000781 Chr.4: 151100942-151100942 on Build GRCh38
rs559283487 Chr.4: 151101048-151101048 on Build GRCh38
rs745484284 Chr.4: 151101114-151101114 on Build GRCh38
rs767951371 Chr.4: 151100950-151100950 on Build GRCh38
rs993185185 Chr.4: 151101087-151101087 on Build GRCh38
rs752291359 Chr.4: 151101028-151101028 on Build GRCh38
rs773937414 Chr.4: 151100956-151100959 on Build GRCh38
rs761149231 Chr.4: 151101135-151101135 on Build GRCh38
rs765469368 Chr.4: 151101003-151101003 on Build GRCh38
rs533195948 Chr.4: 151101077-151101077 on Build GRCh38
rs770298599 Chr.4: 151101096-151101096 on Build GRCh38
rs764663585 Chr.4: 151101137-151101137 on Build GRCh38
rs772610299 Chr.4: 151101127-151101127 on Build GRCh38
rs374926292 Chr.4: 151101072-151101072 on Build GRCh38
rs573414866 Chr.4: 151100969-151100969 on Build GRCh38
rs764654918 Chr.4: 151100966-151100966 on Build GRCh38
rs748844257 Chr.4: 151101092-151101092 on Build GRCh38
rs780944897 Chr.4: 151101070-151101070 on Build GRCh38
rs759138828 Chr.4: 151100958-151100959 on Build GRCh38
rs754809934 Chr.4: 151101069-151101069 on Build GRCh38
rs1025970589 Chr.4: 151101103-151101103 on Build GRCh38
rs776178611 Chr.4: 151100952-151100952 on Build GRCh38
rs761218523 Chr.4: 151100967-151100967 on Build GRCh38
rs1044173367 Chr.4: 151100939-151100939 on Build GRCh38

View Full Product Details