Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 29970879-29971134 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTMR3
Entrez Gene ID
8897
Gene Name
myotubularin related protein 3
Gene Aliases
FYVE-DSP1, ZFYVE10
UniGene
Hs.474536
Gene Chromosome Location
Chr.22: 29883165-30030868 on Build GRCh38
Transcripts
NM_021090.3 NM_153050.2 NM_153051.2 XM_005261803.2 XM_005261804.3 XM_005261805.2 XM_005261806.2 XM_005261808.2 XM_017029028.1 XM_017029029.1 XM_017029030.1 XM_017029031.1 XM_017029032.1 XM_017029034.1

SNP Detail

SNP ID Chromosome Location
rs16988135 Chr.22: 29971025-29971025 on Build GRCh38
rs754782276 Chr.22: 29971016-29971016 on Build GRCh38
rs34749290 Chr.22: 29970935-29970936 on Build GRCh38
rs749681647 Chr.22: 29971070-29971070 on Build GRCh38
rs894067443 Chr.22: 29971090-29971090 on Build GRCh38
rs746428793 Chr.22: 29971042-29971042 on Build GRCh38
rs937232829 Chr.22: 29970915-29970915 on Build GRCh38
rs545862845 Chr.22: 29971085-29971085 on Build GRCh38
rs757325001 Chr.22: 29971027-29971027 on Build GRCh38
rs200732869 Chr.22: 29970946-29970946 on Build GRCh38
rs1056040634 Chr.22: 29970932-29970932 on Build GRCh38
rs772792380 Chr.22: 29971096-29971096 on Build GRCh38
rs10222281 Chr.22: 29970958-29970958 on Build GRCh38
rs538436006 Chr.22: 29971091-29971091 on Build GRCh38
rs751787242 Chr.22: 29971026-29971026 on Build GRCh38
rs577433778 Chr.22: 29971095-29971095 on Build GRCh38
rs949906259 Chr.22: 29970961-29970961 on Build GRCh38
rs778477505 Chr.22: 29971077-29971077 on Build GRCh38
rs769014515 Chr.22: 29971073-29971073 on Build GRCh38
rs752000879 Chr.22: 29971084-29971085 on Build GRCh38
rs199939444 Chr.22: 29971037-29971037 on Build GRCh38
rs753691488 Chr.22: 29971014-29971014 on Build GRCh38
rs6006317 Chr.22: 29970965-29970965 on Build GRCh38
rs749392494 Chr.22: 29970910-29970910 on Build GRCh38
rs756495055 Chr.22: 29971096-29971096 on Build GRCh38
rs372176071 Chr.22: 29971094-29971094 on Build GRCh38
rs866491897 Chr.22: 29970975-29970975 on Build GRCh38
rs370549337 Chr.22: 29971106-29971107 on Build GRCh38
rs199721150 Chr.22: 29971008-29971008 on Build GRCh38
rs1018928283 Chr.22: 29971102-29971102 on Build GRCh38
rs750730990 Chr.22: 29971093-29971096 on Build GRCh38
rs367924231 Chr.22: 29971044-29971044 on Build GRCh38
rs767917449 Chr.22: 29971085-29971086 on Build GRCh38
rs766362513 Chr.22: 29971013-29971013 on Build GRCh38
rs895681268 Chr.22: 29970941-29970941 on Build GRCh38
rs527819555 Chr.22: 29970916-29970916 on Build GRCh38
rs762517321 Chr.22: 29971075-29971075 on Build GRCh38
rs761904634 Chr.22: 29971097-29971100 on Build GRCh38
rs191516634 Chr.22: 29970905-29970905 on Build GRCh38
rs780211488 Chr.22: 29971095-29971096 on Build GRCh38
rs780468936 Chr.22: 29971058-29971058 on Build GRCh38

View Full Product Details