Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 50893467-50893730 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADNP
Entrez Gene ID
23394
Gene Name
activity dependent neuroprotector homeobox
Gene Aliases
ADNP1, HVDAS, MRD28
UniGene
Hs.293736
Gene Chromosome Location
Chr.20: 50888918-50934984 on Build GRCh38
Transcripts
NM_001282531.1 NM_001282532.1 NM_015339.3 NM_181442.2 XM_011528747.2 XM_011528748.2 XM_017027757.1 XM_017027758.1 XM_017027759.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4459097 c.1111C>T p.P371S Chr.20: 50893603-50893603 on Build GRCh38
4458275 c.1079C>T p.P360L Chr.20: 50893635-50893635 on Build GRCh38
444028 c.1041G>A p.M347I Chr.20: 50893673-50893673 on Build GRCh38
1027974 c.1030G>A p.G344S Chr.20: 50893684-50893684 on Build GRCh38
578251 c.1233C>T p.S411S Chr.20: 50893481-50893481 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761934331 Chr.20: 50893577-50893577 on Build GRCh38
rs200625964 Chr.20: 50893651-50893651 on Build GRCh38
rs778671994 Chr.20: 50893603-50893603 on Build GRCh38
rs200732604 Chr.20: 50893551-50893551 on Build GRCh38
rs886056775 Chr.20: 50893612-50893612 on Build GRCh38
rs375916721 Chr.20: 50893633-50893633 on Build GRCh38
rs757949410 Chr.20: 50893607-50893607 on Build GRCh38
rs370787624 Chr.20: 50893592-50893592 on Build GRCh38
rs760486899 Chr.20: 50893587-50893587 on Build GRCh38
rs770111155 Chr.20: 50893580-50893580 on Build GRCh38
rs150949770 Chr.20: 50893680-50893680 on Build GRCh38
rs769876526 Chr.20: 50893640-50893640 on Build GRCh38
rs749861265 Chr.20: 50893606-50893606 on Build GRCh38
rs780570815 Chr.20: 50893536-50893536 on Build GRCh38
rs78300927 Chr.20: 50893639-50893639 on Build GRCh38
rs756841614 Chr.20: 50893602-50893602 on Build GRCh38
rs764261642 Chr.20: 50893499-50893499 on Build GRCh38
rs1051220596 Chr.20: 50893672-50893672 on Build GRCh38
rs772432043 Chr.20: 50893535-50893535 on Build GRCh38
rs769216530 Chr.20: 50893631-50893631 on Build GRCh38
rs201610319 Chr.20: 50893675-50893675 on Build GRCh38
rs756000258 Chr.20: 50893514-50893514 on Build GRCh38
rs747522297 Chr.20: 50893625-50893625 on Build GRCh38
rs763980090 Chr.20: 50893591-50893591 on Build GRCh38
rs1043752877 Chr.20: 50893694-50893694 on Build GRCh38
rs539899954 Chr.20: 50893652-50893652 on Build GRCh38
rs587777524 Chr.20: 50893503-50893503 on Build GRCh38
rs750105424 Chr.20: 50893695-50893695 on Build GRCh38
rs781522818 Chr.20: 50893636-50893636 on Build GRCh38
rs773142534 Chr.20: 50893581-50893581 on Build GRCh38
rs776769119 Chr.20: 50893568-50893568 on Build GRCh38
rs945318495 Chr.20: 50893685-50893685 on Build GRCh38
rs775499611 Chr.20: 50893655-50893655 on Build GRCh38
rs146770784 Chr.20: 50893584-50893584 on Build GRCh38
rs866056890 Chr.20: 50893653-50893653 on Build GRCh38
rs767538651 Chr.20: 50893511-50893511 on Build GRCh38
rs764747481 Chr.20: 50893690-50893690 on Build GRCh38
rs752505309 Chr.20: 50893513-50893513 on Build GRCh38
rs1014650398 Chr.20: 50893572-50893572 on Build GRCh38
rs778276907 Chr.20: 50893530-50893530 on Build GRCh38
rs757004037 Chr.20: 50893524-50893524 on Build GRCh38
rs754857380 Chr.20: 50893510-50893510 on Build GRCh38
rs142624771 Chr.20: 50893666-50893666 on Build GRCh38
rs765870706 Chr.20: 50893700-50893700 on Build GRCh38
rs139951165 Chr.20: 50893502-50893502 on Build GRCh38
rs565881006 Chr.20: 50893648-50893648 on Build GRCh38
rs777516475 Chr.20: 50893515-50893515 on Build GRCh38
rs143474358 Chr.20: 50893534-50893534 on Build GRCh38
rs780483051 Chr.20: 50893621-50893621 on Build GRCh38
rs753551478 Chr.20: 50893518-50893518 on Build GRCh38
rs757837763 Chr.20: 50893696-50893696 on Build GRCh38
rs747807042 Chr.20: 50893642-50893642 on Build GRCh38
rs886039649 Chr.20: 50893606-50893608 on Build GRCh38
rs761702934 Chr.20: 50893683-50893683 on Build GRCh38
rs769022567 Chr.20: 50893567-50893567 on Build GRCh38
rs186717354 Chr.20: 50893586-50893586 on Build GRCh38
rs373919783 Chr.20: 50893495-50893495 on Build GRCh38
rs775813155 Chr.20: 50893493-50893493 on Build GRCh38

View Full Product Details