Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 37643158-37643431 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFP30
Entrez Gene ID
22835
Gene Name
ZFP30 zinc finger protein
Gene Aliases
ZNF745
UniGene
Hs.716719
Gene Chromosome Location
Chr.19: 37632488-37656261 on Build GRCh38
Transcripts
NM_001320666.1 NM_001320667.1 NM_001320668.1 NM_001320669.1 NM_001320670.1 NM_001320671.1 NM_014898.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5613043 c.151T>G p.S51A Chr.19: 37643349-37643349 on Build GRCh38
1393254 c.224G>A p.R75K Chr.19: 37643276-37643276 on Build GRCh38
6151298 c.174A>T p.L58L Chr.19: 37643326-37643326 on Build GRCh38
6151299 c.228G>A p.W76* Chr.19: 37643272-37643272 on Build GRCh38
4077607 c.235+2T>C p.? Chr.19: 37643263-37643263 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746356866 Chr.19: 37643294-37643294 on Build GRCh38
rs545389487 Chr.19: 37643248-37643248 on Build GRCh38
rs761589961 Chr.19: 37643217-37643217 on Build GRCh38
rs117734665 Chr.19: 37643193-37643193 on Build GRCh38
rs200590992 Chr.19: 37643221-37643221 on Build GRCh38
rs752376730 Chr.19: 37643401-37643401 on Build GRCh38
rs766173903 Chr.19: 37643350-37643350 on Build GRCh38
rs980427393 Chr.19: 37643359-37643359 on Build GRCh38
rs201196001 Chr.19: 37643215-37643215 on Build GRCh38
rs750349075 Chr.19: 37643228-37643228 on Build GRCh38
rs762953316 Chr.19: 37643308-37643308 on Build GRCh38
rs753406272 Chr.19: 37643379-37643379 on Build GRCh38
rs760678981 Chr.19: 37643349-37643349 on Build GRCh38
rs779643800 Chr.19: 37643250-37643250 on Build GRCh38
rs781601027 Chr.19: 37643281-37643281 on Build GRCh38
rs1034231966 Chr.19: 37643405-37643405 on Build GRCh38
rs778768597 Chr.19: 37643400-37643400 on Build GRCh38
rs761643145 Chr.19: 37643319-37643319 on Build GRCh38
rs774541292 Chr.19: 37643314-37643314 on Build GRCh38
rs770161747 Chr.19: 37643299-37643299 on Build GRCh38
rs200308197 Chr.19: 37643282-37643282 on Build GRCh38
rs199804131 Chr.19: 37643334-37643334 on Build GRCh38
rs771411767 Chr.19: 37643273-37643273 on Build GRCh38
rs768932455 Chr.19: 37643311-37643311 on Build GRCh38
rs754696249 Chr.19: 37643252-37643252 on Build GRCh38
rs1005581875 Chr.19: 37643201-37643203 on Build GRCh38
rs750142776 Chr.19: 37643348-37643348 on Build GRCh38
rs754534743 Chr.19: 37643396-37643396 on Build GRCh38
rs1017343175 Chr.19: 37643274-37643274 on Build GRCh38
rs1028831940 Chr.19: 37643342-37643342 on Build GRCh38
rs774419948 Chr.19: 37643271-37643272 on Build GRCh38
rs767309679 Chr.19: 37643327-37643327 on Build GRCh38
rs753346679 Chr.19: 37643251-37643251 on Build GRCh38
rs751313129 Chr.19: 37643305-37643305 on Build GRCh38
rs778645931 Chr.19: 37643266-37643266 on Build GRCh38
rs747447991 Chr.19: 37643269-37643269 on Build GRCh38

View Full Product Details