Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 21426424-21426697 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNKSR2
Entrez Gene ID
22866
Gene Name
connector enhancer of kinase suppressor of Ras 2
Gene Aliases
CNK2, KSR2, MAGUIN
UniGene
Hs.555917
Gene Chromosome Location
Chr.X: 21374335-21654695 on Build GRCh38
Transcripts
NM_001168647.1 NM_001168648.1 NM_001168649.1 NM_014927.3 XM_011545471.2 XM_011545472.2 XM_017029358.1

SNP Detail

SNP ID Chromosome Location
rs771705122 Chr.X: 21426569-21426569 on Build GRCh38
rs767663747 Chr.X: 21426552-21426552 on Build GRCh38
rs963671924 Chr.X: 21426488-21426488 on Build GRCh38
rs557588406 Chr.X: 21426669-21426669 on Build GRCh38
rs779203031 Chr.X: 21426627-21426627 on Build GRCh38
rs757776373 Chr.X: 21426624-21426624 on Build GRCh38
rs749730618 Chr.X: 21426619-21426619 on Build GRCh38
rs886041712 Chr.X: 21426546-21426546 on Build GRCh38
rs776485195 Chr.X: 21426512-21426512 on Build GRCh38
rs752477728 Chr.X: 21426534-21426534 on Build GRCh38
rs750254579 Chr.X: 21426536-21426536 on Build GRCh38
rs760146378 Chr.X: 21426461-21426461 on Build GRCh38
rs755200292 Chr.X: 21426540-21426540 on Build GRCh38
rs895454787 Chr.X: 21426607-21426607 on Build GRCh38
rs761693560 Chr.X: 21426522-21426522 on Build GRCh38
rs756206673 Chr.X: 21426568-21426568 on Build GRCh38
rs764065873 Chr.X: 21426504-21426504 on Build GRCh38
rs113885403 Chr.X: 21426630-21426630 on Build GRCh38
rs752790380 Chr.X: 21426564-21426564 on Build GRCh38

View Full Product Details