Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 5634712-5634939 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIM34
Entrez Gene ID
53840
Gene Name
tripartite motif containing 34
Gene Aliases
IFP1, RNF21
UniGene
Hs.729048
Gene Chromosome Location
Chr.11: 5619944-5644398 on Build GRCh38
Transcripts
NM_001003827.1 NM_021616.5 NM_130390.2 NM_001003819.3

Gene Symbol
TRIM6-TRIM34
Entrez Gene ID
445372
Gene Name
TRIM6-TRIM34 readthrough
Gene Aliases
IFP1, RNF21, TRIM34
UniGene
Hs.729048
Gene Chromosome Location
Chr.11: 5596635-5644398 on Build GRCh38
Transcripts
NM_001003827.1 NM_021616.5 NM_130390.2 NM_001003819.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3397864 c.1758G>T p.E586D Chr.11: 5634807-5634807 on Build GRCh38
928489 c.635C>T p.T212M Chr.11: 5634746-5634746 on Build GRCh38
5035060 c.1740G>A p.Q580Q Chr.11: 5634789-5634789 on Build GRCh38
928488 c.1697C>T p.T566M Chr.11: 5634746-5634746 on Build GRCh38
5035061 c.678G>A p.Q226Q Chr.11: 5634789-5634789 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs61758098 Chr.11: 5634758-5634758 on Build GRCh38
rs756928577 Chr.11: 5634835-5634835 on Build GRCh38
rs1025337033 Chr.11: 5634736-5634736 on Build GRCh38
rs760063922 Chr.11: 5634910-5634910 on Build GRCh38
rs776593790 Chr.11: 5634798-5634798 on Build GRCh38
rs189759419 Chr.11: 5634894-5634894 on Build GRCh38
rs750770702 Chr.11: 5634739-5634739 on Build GRCh38
rs769915606 Chr.11: 5634811-5634811 on Build GRCh38
rs528924303 Chr.11: 5634752-5634752 on Build GRCh38
rs750209956 Chr.11: 5634848-5634848 on Build GRCh38
rs780014650 Chr.11: 5634795-5634795 on Build GRCh38
rs376140381 Chr.11: 5634747-5634747 on Build GRCh38
rs200628102 Chr.11: 5634797-5634797 on Build GRCh38
rs939309979 Chr.11: 5634871-5634871 on Build GRCh38
rs927540880 Chr.11: 5634807-5634807 on Build GRCh38
rs374915648 Chr.11: 5634906-5634906 on Build GRCh38
rs991942139 Chr.11: 5634754-5634754 on Build GRCh38
rs745714767 Chr.11: 5634774-5634774 on Build GRCh38
rs758228221 Chr.11: 5634858-5634858 on Build GRCh38
rs760151929 Chr.11: 5634829-5634829 on Build GRCh38
rs557049718 Chr.11: 5634918-5634918 on Build GRCh38
rs139090035 Chr.11: 5634746-5634746 on Build GRCh38
rs898624962 Chr.11: 5634919-5634919 on Build GRCh38
rs553635308 Chr.11: 5634883-5634883 on Build GRCh38
rs779852754 Chr.11: 5634870-5634870 on Build GRCh38
rs770047012 Chr.11: 5634885-5634885 on Build GRCh38
rs372510799 Chr.11: 5634909-5634909 on Build GRCh38
rs371652236 Chr.11: 5634879-5634879 on Build GRCh38
rs117087264 Chr.11: 5634874-5634874 on Build GRCh38
rs980684124 Chr.11: 5634794-5634794 on Build GRCh38
rs749640906 Chr.11: 5634899-5634899 on Build GRCh38
rs778700935 Chr.11: 5634762-5634762 on Build GRCh38
rs145787574 Chr.11: 5634830-5634830 on Build GRCh38
rs753491596 Chr.11: 5634755-5634755 on Build GRCh38
rs763453391 Chr.11: 5634741-5634741 on Build GRCh38
rs764853741 Chr.11: 5634844-5634844 on Build GRCh38
rs753302972 Chr.11: 5634831-5634831 on Build GRCh38
rs535888165 Chr.11: 5634907-5634907 on Build GRCh38
rs151152683 Chr.11: 5634745-5634745 on Build GRCh38
rs773624006 Chr.11: 5634817-5634817 on Build GRCh38
rs147047595 Chr.11: 5634827-5634827 on Build GRCh38
rs371997677 Chr.11: 5634824-5634824 on Build GRCh38
rs199617591 Chr.11: 5634873-5634873 on Build GRCh38
rs747056223 Chr.11: 5634796-5634796 on Build GRCh38
rs762033958 Chr.11: 5634802-5634802 on Build GRCh38
rs771984479 Chr.11: 5634789-5634789 on Build GRCh38

View Full Product Details