Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 12260699-12260956 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS13D
Entrez Gene ID
55187
Gene Name
vacuolar protein sorting 13 homolog D
Gene Aliases
-
UniGene
Hs.439381
Gene Chromosome Location
Chr.1: 12230039-12512047 on Build GRCh38
Transcripts
NM_015378.3 NM_018156.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4522124 c.1147G>A p.E383K Chr.1: 12260729-12260729 on Build GRCh38
5804149 c.1133A>T p.E378V Chr.1: 12260715-12260715 on Build GRCh38
894803 c.1124G>A p.R375Q Chr.1: 12260706-12260706 on Build GRCh38
894802 c.1123C>T p.R375W Chr.1: 12260705-12260705 on Build GRCh38
1687016 c.1183C>T p.R395* Chr.1: 12260765-12260765 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs6657877 Chr.1: 12260892-12260892 on Build GRCh38
rs1017669888 Chr.1: 12260729-12260729 on Build GRCh38
rs768737006 Chr.1: 12260747-12260747 on Build GRCh38
rs996685089 Chr.1: 12260756-12260761 on Build GRCh38
rs752015762 Chr.1: 12260784-12260784 on Build GRCh38
rs555141309 Chr.1: 12260886-12260886 on Build GRCh38
rs750321685 Chr.1: 12260766-12260766 on Build GRCh38
rs985951184 Chr.1: 12260927-12260927 on Build GRCh38
rs774379629 Chr.1: 12260898-12260898 on Build GRCh38
rs776442029 Chr.1: 12260920-12260920 on Build GRCh38
rs747979971 Chr.1: 12260906-12260906 on Build GRCh38
rs757502708 Chr.1: 12260787-12260787 on Build GRCh38
rs765941549 Chr.1: 12260774-12260774 on Build GRCh38
rs760920787 Chr.1: 12260843-12260843 on Build GRCh38
rs138147813 Chr.1: 12260737-12260737 on Build GRCh38
rs375214360 Chr.1: 12260923-12260923 on Build GRCh38
rs752359125 Chr.1: 12260765-12260765 on Build GRCh38
rs12033054 Chr.1: 12260807-12260807 on Build GRCh38
rs149554788 Chr.1: 12260748-12260748 on Build GRCh38
rs781723691 Chr.1: 12260802-12260802 on Build GRCh38
rs760294849 Chr.1: 12260769-12260769 on Build GRCh38
rs749334750 Chr.1: 12260838-12260838 on Build GRCh38
rs775717158 Chr.1: 12260736-12260736 on Build GRCh38
rs905420307 Chr.1: 12260790-12260790 on Build GRCh38
rs767503767 Chr.1: 12260756-12260756 on Build GRCh38
rs1003976998 Chr.1: 12260808-12260808 on Build GRCh38
rs1022348390 Chr.1: 12260887-12260887 on Build GRCh38
rs778430207 Chr.1: 12260783-12260783 on Build GRCh38
rs370704795 Chr.1: 12260778-12260778 on Build GRCh38
rs769883435 Chr.1: 12260835-12260835 on Build GRCh38
rs535567591 Chr.1: 12260836-12260836 on Build GRCh38

View Full Product Details