Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154235109-154235356 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBAP2L
Entrez Gene ID
9898
Gene Name
ubiquitin associated protein 2 like
Gene Aliases
NICE-4, NICE4
UniGene
Hs.490551
Gene Chromosome Location
Chr.1: 154220172-154271510 on Build GRCh38
Transcripts
NM_001127320.2 NM_001287815.1 NM_001287816.1 NM_014847.3 XM_005245658.3 XM_005245667.3 XM_005245668.3 XM_005245669.3 XM_005245670.3 XM_005245672.3 XM_005245673.3 XM_005245674.1 XM_011510215.2 XM_011510216.1 XM_017002972.1 XM_017002973.1 XM_017002974.1 XM_017002975.1 XM_017002976.1 XM_017002977.1 XM_017002978.1 XM_017002979.1 XM_017002980.1 XM_017002981.1 XM_017002982.1 XM_017002983.1 XM_017002984.1 XM_017002985.1 XM_017002986.1 XM_017002987.1 XM_017002988.1 XM_017002989.1 XM_017002990.1 XM_017002991.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4221832 c.492G>T p.Q164H Chr.1: 154235206-154235206 on Build GRCh38
2208684 c.459G>T p.Q153H Chr.1: 154235206-154235206 on Build GRCh38
3979850 c.453A>T p.R151R Chr.1: 154235200-154235200 on Build GRCh38
1747781 c.472G>A p.D158N Chr.1: 154235219-154235219 on Build GRCh38
4140846 c.524G>T p.G175V Chr.1: 154235238-154235238 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576151360 Chr.1: 154235281-154235281 on Build GRCh38
rs557688567 Chr.1: 154235241-154235241 on Build GRCh38
rs757153961 Chr.1: 154235174-154235174 on Build GRCh38
rs148736933 Chr.1: 154235287-154235287 on Build GRCh38
rs779781450 Chr.1: 154235324-154235324 on Build GRCh38
rs763416460 Chr.1: 154235253-154235253 on Build GRCh38
rs545838756 Chr.1: 154235165-154235165 on Build GRCh38
rs762240663 Chr.1: 154235240-154235240 on Build GRCh38
rs766690379 Chr.1: 154235151-154235151 on Build GRCh38
rs1043634608 Chr.1: 154235216-154235216 on Build GRCh38
rs1020720230 Chr.1: 154235202-154235202 on Build GRCh38
rs201031131 Chr.1: 154235151-154235152 on Build GRCh38
rs572056717 Chr.1: 154235161-154235161 on Build GRCh38
rs12023901 Chr.1: 154235159-154235159 on Build GRCh38
rs113785380 Chr.1: 154235195-154235195 on Build GRCh38
rs745765867 Chr.1: 154235311-154235311 on Build GRCh38
rs751854018 Chr.1: 154235277-154235277 on Build GRCh38
rs752770977 Chr.1: 154235289-154235289 on Build GRCh38
rs942237274 Chr.1: 154235152-154235152 on Build GRCh38
rs762628285 Chr.1: 154235143-154235143 on Build GRCh38
rs1056473986 Chr.1: 154235251-154235251 on Build GRCh38
rs779484404 Chr.1: 154235159-154235159 on Build GRCh38
rs532984518 Chr.1: 154235156-154235157 on Build GRCh38
rs369112672 Chr.1: 154235173-154235174 on Build GRCh38
rs755478359 Chr.1: 154235150-154235151 on Build GRCh38
rs144300846 Chr.1: 154235199-154235199 on Build GRCh38
rs375690428 Chr.1: 154235225-154235225 on Build GRCh38
rs576757218 Chr.1: 154235156-154235156 on Build GRCh38
rs756138799 Chr.1: 154235303-154235303 on Build GRCh38
rs778589980 Chr.1: 154235176-154235176 on Build GRCh38
rs567039769 Chr.1: 154235157-154235159 on Build GRCh38
rs746613043 Chr.1: 154235203-154235203 on Build GRCh38
rs745737509 Chr.1: 154235187-154235187 on Build GRCh38
rs1012329877 Chr.1: 154235283-154235283 on Build GRCh38
rs746667993 Chr.1: 154235326-154235326 on Build GRCh38
rs771729624 Chr.1: 154235198-154235198 on Build GRCh38
rs776050263 Chr.1: 154235233-154235233 on Build GRCh38
rs778946004 Chr.1: 154235310-154235310 on Build GRCh38
rs766830634 Chr.1: 154235263-154235263 on Build GRCh38
rs774040771 Chr.1: 154235318-154235318 on Build GRCh38
rs779985986 Chr.1: 154235223-154235223 on Build GRCh38
rs865865980 Chr.1: 154235228-154235228 on Build GRCh38

View Full Product Details