Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 12343012-12343273 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VWDE
Entrez Gene ID
221806
Gene Name
von Willebrand factor D and EGF domains
Gene Aliases
-
UniGene
Hs.669526
Gene Chromosome Location
Chr.7: 12330883-12404226 on Build GRCh38
Transcripts
NM_001135924.1 XM_005249641.1 XM_011515174.1 XM_011515175.1 XM_017011828.1 XM_017011829.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4493387 c.4142C>T p.P1381L Chr.7: 12343115-12343115 on Build GRCh38
4493385 c.4141C>T p.P1381S Chr.7: 12343116-12343116 on Build GRCh38
6548004 c.4159C>A p.P1387T Chr.7: 12343098-12343098 on Build GRCh38
5668313 c.4119G>T p.L1373F Chr.7: 12343138-12343138 on Build GRCh38
5668314 c.1896G>T p.L632F Chr.7: 12343138-12343138 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199627971 Chr.7: 12343144-12343144 on Build GRCh38
rs953941135 Chr.7: 12343185-12343185 on Build GRCh38
rs567216138 Chr.7: 12343180-12343180 on Build GRCh38
rs776364227 Chr.7: 12343103-12343103 on Build GRCh38
rs968291560 Chr.7: 12343221-12343221 on Build GRCh38
rs752332677 Chr.7: 12343219-12343219 on Build GRCh38
rs374183450 Chr.7: 12343110-12343110 on Build GRCh38
rs761999599 Chr.7: 12343130-12343130 on Build GRCh38
rs919666409 Chr.7: 12343052-12343052 on Build GRCh38
rs1029309699 Chr.7: 12343148-12343148 on Build GRCh38
rs867554346 Chr.7: 12343065-12343065 on Build GRCh38
rs905158625 Chr.7: 12343143-12343143 on Build GRCh38
rs1044955849 Chr.7: 12343111-12343111 on Build GRCh38
rs766969427 Chr.7: 12343181-12343181 on Build GRCh38
rs779199368 Chr.7: 12343217-12343217 on Build GRCh38
rs977992969 Chr.7: 12343225-12343225 on Build GRCh38
rs1030418523 Chr.7: 12343232-12343232 on Build GRCh38
rs555788673 Chr.7: 12343237-12343237 on Build GRCh38
rs917546360 Chr.7: 12343104-12343104 on Build GRCh38
rs770883093 Chr.7: 12343055-12343055 on Build GRCh38
rs369817888 Chr.7: 12343109-12343109 on Build GRCh38
rs534514042 Chr.7: 12343230-12343235 on Build GRCh38
rs915042277 Chr.7: 12343211-12343211 on Build GRCh38
rs755364492 Chr.7: 12343195-12343195 on Build GRCh38
rs890427382 Chr.7: 12343234-12343234 on Build GRCh38
rs1046831055 Chr.7: 12343093-12343093 on Build GRCh38
rs549834306 Chr.7: 12343168-12343168 on Build GRCh38
rs113602532 Chr.7: 12343229-12343229 on Build GRCh38
rs375861928 Chr.7: 12343045-12343045 on Build GRCh38
rs754434958 Chr.7: 12343188-12343188 on Build GRCh38

View Full Product Details