Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 209608887-209609154 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMK1G
Entrez Gene ID
57172
Gene Name
calcium/calmodulin dependent protein kinase IG
Gene Aliases
CLICK3, CLICKIII, VWS1, dJ272L16.1
UniGene
Hs.199068
Gene Chromosome Location
Chr.1: 209583700-209613939 on Build GRCh38
Transcripts
NM_020439.2 XM_017001866.1 XM_017001867.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4429093 c.733G>A p.D245N Chr.1: 209609077-209609077 on Build GRCh38
1748126 c.746C>T p.S249L Chr.1: 209609090-209609090 on Build GRCh38
6619173 c.648delC p.P218fs*55 Chr.1: 209608992-209608992 on Build GRCh38
6619175 c.644G>A p.G215E Chr.1: 209608988-209608988 on Build GRCh38
4504004 c.653C>T p.P218L Chr.1: 209608997-209608997 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762140202 Chr.1: 209609047-209609047 on Build GRCh38
rs765752817 Chr.1: 209609086-209609086 on Build GRCh38
rs764308165 Chr.1: 209608929-209608929 on Build GRCh38
rs144298174 Chr.1: 209608992-209608992 on Build GRCh38
rs149003369 Chr.1: 209609028-209609028 on Build GRCh38
rs746018496 Chr.1: 209609107-209609107 on Build GRCh38
rs11119314 Chr.1: 209608998-209608998 on Build GRCh38
rs779846053 Chr.1: 209608910-209608910 on Build GRCh38
rs537213845 Chr.1: 209609052-209609052 on Build GRCh38
rs779956913 Chr.1: 209609119-209609119 on Build GRCh38
rs368036491 Chr.1: 209608968-209608968 on Build GRCh38
rs772934563 Chr.1: 209609130-209609130 on Build GRCh38
rs199812064 Chr.1: 209609012-209609012 on Build GRCh38
rs765147315 Chr.1: 209609096-209609096 on Build GRCh38
rs747266239 Chr.1: 209608997-209608997 on Build GRCh38
rs573839493 Chr.1: 209609108-209609108 on Build GRCh38
rs774645531 Chr.1: 209609030-209609030 on Build GRCh38
rs1043440645 Chr.1: 209608925-209608925 on Build GRCh38
rs776077217 Chr.1: 209609018-209609018 on Build GRCh38
rs781544486 Chr.1: 209609004-209609004 on Build GRCh38
rs749981369 Chr.1: 209608965-209608965 on Build GRCh38
rs749287633 Chr.1: 209608970-209608970 on Build GRCh38
rs748577038 Chr.1: 209609126-209609126 on Build GRCh38
rs754831027 Chr.1: 209609093-209609093 on Build GRCh38
rs749820011 Chr.1: 209609121-209609121 on Build GRCh38
rs760294215 Chr.1: 209609081-209609081 on Build GRCh38
rs780108701 Chr.1: 209608985-209608985 on Build GRCh38
rs772850104 Chr.1: 209609064-209609064 on Build GRCh38
rs758238601 Chr.1: 209609101-209609101 on Build GRCh38
rs761411304 Chr.1: 209608945-209608945 on Build GRCh38
rs1016792476 Chr.1: 209609040-209609040 on Build GRCh38
rs781620827 Chr.1: 209609104-209609104 on Build GRCh38
rs769092278 Chr.1: 209609029-209609029 on Build GRCh38
rs191863683 Chr.1: 209609088-209609088 on Build GRCh38
rs774805053 Chr.1: 209609125-209609125 on Build GRCh38
rs372047332 Chr.1: 209608996-209608996 on Build GRCh38
rs947729068 Chr.1: 209608922-209608922 on Build GRCh38
rs1019025157 Chr.1: 209609069-209609069 on Build GRCh38
rs147278175 Chr.1: 209609013-209609013 on Build GRCh38
rs767195803 Chr.1: 209609048-209609048 on Build GRCh38
rs567514631 Chr.1: 209609073-209609073 on Build GRCh38
rs878869929 Chr.1: 209609075-209609075 on Build GRCh38
rs767032560 Chr.1: 209608958-209608958 on Build GRCh38
rs756222267 Chr.1: 209609112-209609112 on Build GRCh38
rs373764707 Chr.1: 209609100-209609100 on Build GRCh38
rs895861870 Chr.1: 209608995-209608995 on Build GRCh38
rs769196234 Chr.1: 209609123-209609123 on Build GRCh38
rs372730608 Chr.1: 209608935-209608935 on Build GRCh38

View Full Product Details