Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 87885523-87885794 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC497256
Entrez Gene ID
497256
Gene Name
uncharacterized LOC497256
Gene Aliases
-
UniGene
Hs.584108
Gene Chromosome Location
Chr.9: 87844517-87939244 on Build GRCh38
Transcripts
NM_178828.4

Gene Symbol
SPATA31E1
Entrez Gene ID
286234
Gene Name
SPATA31 subfamily E member 1
Gene Aliases
C9orf79, FAM75E1
UniGene
Hs.130672
Gene Chromosome Location
Chr.9: 87882857-87888899 on Build GRCh38
Transcripts
NM_178828.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3659351 c.1213C>T p.P405S Chr.9: 87885700-87885700 on Build GRCh38
1555669 c.1154G>A p.R385Q Chr.9: 87885641-87885641 on Build GRCh38
3728416 c.1166C>A p.P389Q Chr.9: 87885653-87885653 on Build GRCh38
3659352 c.1214C>T p.P405L Chr.9: 87885701-87885701 on Build GRCh38
6184483 c.1202C>T p.T401M Chr.9: 87885689-87885689 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs34946554 Chr.9: 87885712-87885712 on Build GRCh38
rs1041531692 Chr.9: 87885657-87885657 on Build GRCh38
rs764330261 Chr.9: 87885587-87885587 on Build GRCh38
rs747657173 Chr.9: 87885609-87885609 on Build GRCh38
rs200696251 Chr.9: 87885677-87885677 on Build GRCh38
rs556963122 Chr.9: 87885690-87885690 on Build GRCh38
rs368642338 Chr.9: 87885769-87885769 on Build GRCh38
rs761255850 Chr.9: 87885584-87885585 on Build GRCh38
rs752185075 Chr.9: 87885663-87885663 on Build GRCh38
rs768027216 Chr.9: 87885638-87885638 on Build GRCh38
rs62578207 Chr.9: 87885555-87885555 on Build GRCh38
rs781138546 Chr.9: 87885660-87885660 on Build GRCh38
rs753359007 Chr.9: 87885773-87885773 on Build GRCh38
rs534832110 Chr.9: 87885595-87885595 on Build GRCh38
rs565943566 Chr.9: 87885568-87885568 on Build GRCh38
rs756146871 Chr.9: 87885626-87885629 on Build GRCh38
rs754891073 Chr.9: 87885602-87885602 on Build GRCh38
rs573180007 Chr.9: 87885740-87885740 on Build GRCh38
rs758158604 Chr.9: 87885594-87885594 on Build GRCh38
rs750300356 Chr.9: 87885608-87885608 on Build GRCh38
rs755642872 Chr.9: 87885668-87885668 on Build GRCh38
rs201074253 Chr.9: 87885768-87885768 on Build GRCh38
rs984119457 Chr.9: 87885645-87885645 on Build GRCh38
rs748958456 Chr.9: 87885680-87885680 on Build GRCh38
rs759964551 Chr.9: 87885628-87885628 on Build GRCh38
rs768709763 Chr.9: 87885711-87885711 on Build GRCh38
rs559642473 Chr.9: 87885739-87885739 on Build GRCh38
rs75372993 Chr.9: 87885626-87885626 on Build GRCh38
rs762031309 Chr.9: 87885718-87885718 on Build GRCh38
rs754055214 Chr.9: 87885642-87885642 on Build GRCh38
rs779450573 Chr.9: 87885689-87885689 on Build GRCh38
rs780574227 Chr.9: 87885606-87885606 on Build GRCh38
rs758248370 Chr.9: 87885643-87885643 on Build GRCh38
rs777622737 Chr.9: 87885679-87885679 on Build GRCh38
rs746939917 Chr.9: 87885596-87885596 on Build GRCh38
rs536605501 Chr.9: 87885552-87885552 on Build GRCh38
rs769378792 Chr.9: 87885611-87885611 on Build GRCh38
rs752663485 Chr.9: 87885756-87885756 on Build GRCh38
rs140330362 Chr.9: 87885607-87885607 on Build GRCh38
rs752755586 Chr.9: 87885571-87885571 on Build GRCh38
rs367546357 Chr.9: 87885573-87885573 on Build GRCh38
rs760733880 Chr.9: 87885703-87885703 on Build GRCh38
rs199568188 Chr.9: 87885567-87885567 on Build GRCh38
rs201760978 Chr.9: 87885627-87885627 on Build GRCh38
rs202211642 Chr.9: 87885646-87885646 on Build GRCh38
rs200735015 Chr.9: 87885551-87885551 on Build GRCh38
rs751293516 Chr.9: 87885653-87885653 on Build GRCh38
rs745604223 Chr.9: 87885772-87885772 on Build GRCh38
rs767436261 Chr.9: 87885605-87885606 on Build GRCh38
rs201505051 Chr.9: 87885641-87885641 on Build GRCh38
rs372761849 Chr.9: 87885554-87885554 on Build GRCh38
rs145538284 Chr.9: 87885694-87885694 on Build GRCh38
rs774990542 Chr.9: 87885557-87885557 on Build GRCh38
rs765405705 Chr.9: 87885738-87885738 on Build GRCh38
rs772505474 Chr.9: 87885693-87885693 on Build GRCh38
rs772966146 Chr.9: 87885620-87885620 on Build GRCh38
rs199958961 Chr.9: 87885640-87885640 on Build GRCh38
rs118103831 Chr.9: 87885654-87885654 on Build GRCh38
rs770639369 Chr.9: 87885682-87885682 on Build GRCh38

View Full Product Details