Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 63169272-63169545 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YTHDF3
Entrez Gene ID
253943
Gene Name
YTH N6-methyladenosine RNA binding protein 3
Gene Aliases
-
UniGene
Hs.491861
Gene Chromosome Location
Chr.8: 63168553-63212788 on Build GRCh38
Transcripts
NM_001277813.1 NM_001277814.1 NM_001277815.1 NM_001277816.1 NM_001277817.1 NM_001277818.1 NM_152758.5 NR_102434.1 XM_011517509.2 XM_011517510.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1624116 c.49+9G>A p.? Chr.8: 63169420-63169420 on Build GRCh38
6472656 c.39A>G p.Q13Q Chr.8: 63169401-63169401 on Build GRCh38
4138725 c.32A>G p.K11R Chr.8: 63169394-63169394 on Build GRCh38
7242835 c.25-1G>A p.? Chr.8: 63169386-63169386 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs764964182 Chr.8: 63169342-63169342 on Build GRCh38
rs368771863 Chr.8: 63169380-63169380 on Build GRCh38
rs993822669 Chr.8: 63169512-63169512 on Build GRCh38
rs759847193 Chr.8: 63169345-63169345 on Build GRCh38
rs757583548 Chr.8: 63169366-63169366 on Build GRCh38
rs746308100 Chr.8: 63169372-63169372 on Build GRCh38
rs370787416 Chr.8: 63169445-63169445 on Build GRCh38
rs760783595 Chr.8: 63169446-63169446 on Build GRCh38
rs901414778 Chr.8: 63169465-63169465 on Build GRCh38
rs751128434 Chr.8: 63169352-63169354 on Build GRCh38
rs889551784 Chr.8: 63169478-63169478 on Build GRCh38
rs775098142 Chr.8: 63169375-63169375 on Build GRCh38
rs374996083 Chr.8: 63169473-63169473 on Build GRCh38
rs779150252 Chr.8: 63169368-63169368 on Build GRCh38
rs779274870 Chr.8: 63169333-63169333 on Build GRCh38
rs769568813 Chr.8: 63169404-63169404 on Build GRCh38
rs761629188 Chr.8: 63169340-63169340 on Build GRCh38
rs780928122 Chr.8: 63169422-63169423 on Build GRCh38
rs1005675511 Chr.8: 63169480-63169480 on Build GRCh38
rs970371353 Chr.8: 63169399-63169399 on Build GRCh38
rs763509289 Chr.8: 63169342-63169343 on Build GRCh38
rs878970538 Chr.8: 63169462-63169462 on Build GRCh38
rs1016619213 Chr.8: 63169504-63169504 on Build GRCh38
rs755912619 Chr.8: 63169434-63169434 on Build GRCh38
rs917102417 Chr.8: 63169332-63169332 on Build GRCh38
rs980465377 Chr.8: 63169472-63169472 on Build GRCh38
rs535408667 Chr.8: 63169509-63169510 on Build GRCh38
rs778157699 Chr.8: 63169315-63169315 on Build GRCh38
rs768185920 Chr.8: 63169390-63169390 on Build GRCh38
rs747076123 Chr.8: 63169317-63169317 on Build GRCh38
rs771201488 Chr.8: 63169330-63169330 on Build GRCh38
rs539082931 Chr.8: 63169476-63169476 on Build GRCh38
rs754319952 Chr.8: 63169363-63169363 on Build GRCh38
rs989251544 Chr.8: 63169510-63169510 on Build GRCh38
rs957734036 Chr.8: 63169479-63169479 on Build GRCh38
rs374501062 Chr.8: 63169321-63169321 on Build GRCh38
rs971250700 Chr.8: 63169337-63169337 on Build GRCh38
rs748052481 Chr.8: 63169401-63169401 on Build GRCh38
rs745595473 Chr.8: 63169423-63169425 on Build GRCh38
rs752814057 Chr.8: 63169352-63169352 on Build GRCh38
rs756299619 Chr.8: 63169359-63169359 on Build GRCh38
rs759034117 Chr.8: 63169342-63169345 on Build GRCh38
rs75630201 Chr.8: 63169362-63169362 on Build GRCh38
rs994885261 Chr.8: 63169331-63169331 on Build GRCh38
rs1007539082 Chr.8: 63169303-63169303 on Build GRCh38
rs542699679 Chr.8: 63169438-63169438 on Build GRCh38
rs1024917698 Chr.8: 63169388-63169388 on Build GRCh38
rs1028096420 Chr.8: 63169312-63169312 on Build GRCh38
rs767920575 Chr.8: 63169350-63169350 on Build GRCh38
rs772881267 Chr.8: 63169406-63169406 on Build GRCh38
rs781136452 Chr.8: 63169397-63169397 on Build GRCh38
rs756809083 Chr.8: 63169423-63169423 on Build GRCh38

View Full Product Details