Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 111343986-111344259 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TEX29
Entrez Gene ID
121793
Gene Name
testis expressed 29
Gene Aliases
C13orf16, bA474D23.1
UniGene
Hs.210677
Gene Chromosome Location
Chr.13: 111305838-111344248 on Build GRCh38
Transcripts
NM_001303133.1 NM_152324.2 XM_017020387.1 XM_017020388.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5043595 c.453C>A p.D151E Chr.13: 111344120-111344120 on Build GRCh38
3671237 c.439G>A p.E147K Chr.13: 111344106-111344106 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs907996033 Chr.13: 111344014-111344014 on Build GRCh38
rs780224800 Chr.13: 111344135-111344137 on Build GRCh38
rs769134877 Chr.13: 111344141-111344141 on Build GRCh38
rs76083530 Chr.13: 111344201-111344201 on Build GRCh38
rs574861359 Chr.13: 111344026-111344026 on Build GRCh38
rs762650326 Chr.13: 111344053-111344053 on Build GRCh38
rs759263382 Chr.13: 111344175-111344175 on Build GRCh38
rs772701280 Chr.13: 111344042-111344042 on Build GRCh38
rs34062346 Chr.13: 111344127-111344128 on Build GRCh38
rs777311457 Chr.13: 111344165-111344165 on Build GRCh38
rs759334392 Chr.13: 111344076-111344076 on Build GRCh38
rs767379293 Chr.13: 111344081-111344081 on Build GRCh38
rs752622910 Chr.13: 111344086-111344086 on Build GRCh38
rs560755834 Chr.13: 111344106-111344106 on Build GRCh38
rs963472338 Chr.13: 111344222-111344222 on Build GRCh38
rs1039016086 Chr.13: 111344206-111344206 on Build GRCh38
rs997603035 Chr.13: 111344066-111344066 on Build GRCh38
rs774054539 Chr.13: 111344063-111344063 on Build GRCh38
rs747615989 Chr.13: 111344140-111344140 on Build GRCh38
rs1026492722 Chr.13: 111344099-111344099 on Build GRCh38
rs116905328 Chr.13: 111344172-111344172 on Build GRCh38
rs201524207 Chr.13: 111344028-111344028 on Build GRCh38
rs758504681 Chr.13: 111344061-111344063 on Build GRCh38
rs112999083 Chr.13: 111344130-111344130 on Build GRCh38
rs111680027 Chr.13: 111344221-111344221 on Build GRCh38
rs779250820 Chr.13: 111344119-111344119 on Build GRCh38
rs951003067 Chr.13: 111344104-111344104 on Build GRCh38
rs973490954 Chr.13: 111344230-111344230 on Build GRCh38
rs921929648 Chr.13: 111344051-111344051 on Build GRCh38
rs1005079618 Chr.13: 111344214-111344214 on Build GRCh38
rs114872122 Chr.13: 111344166-111344166 on Build GRCh38
rs889735020 Chr.13: 111344184-111344184 on Build GRCh38
rs372923307 Chr.13: 111344058-111344058 on Build GRCh38
rs985076336 Chr.13: 111344105-111344105 on Build GRCh38
rs200134927 Chr.13: 111344131-111344131 on Build GRCh38
rs781551813 Chr.13: 111344117-111344117 on Build GRCh38
rs558461812 Chr.13: 111344202-111344202 on Build GRCh38
rs150151490 Chr.13: 111344133-111344133 on Build GRCh38
rs368660128 Chr.13: 111344040-111344040 on Build GRCh38
rs760844578 Chr.13: 111344096-111344096 on Build GRCh38

View Full Product Details