Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 43079499-43079754 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POMGNT2
Entrez Gene ID
84892
Gene Name
protein O-linked mannose N-acetylglucosaminyltransferase 2 (beta 1,4-)
Gene Aliases
AGO61, C3orf39, GTDC2, MDDGA8
UniGene
Hs.745002
Gene Chromosome Location
Chr.3: 43079229-43106092 on Build GRCh38
Transcripts
NM_032806.5 XM_005265515.2 XM_011534163.2 XM_011534164.1 XM_017007353.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1186250 c.1729G>T p.V577L Chr.3: 43079703-43079703 on Build GRCh38
396037 c.1733G>A p.C578Y Chr.3: 43079699-43079699 on Build GRCh38
6676665 c.1681C>T p.R561C Chr.3: 43079751-43079751 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs370391130 Chr.3: 43079649-43079649 on Build GRCh38
rs560585843 Chr.3: 43079641-43079641 on Build GRCh38
rs575514680 Chr.3: 43079563-43079563 on Build GRCh38
rs539221096 Chr.3: 43079549-43079549 on Build GRCh38
rs766377193 Chr.3: 43079666-43079666 on Build GRCh38
rs960690723 Chr.3: 43079556-43079556 on Build GRCh38
rs1035263747 Chr.3: 43079553-43079553 on Build GRCh38
rs532544685 Chr.3: 43079680-43079680 on Build GRCh38
rs1053325191 Chr.3: 43079660-43079660 on Build GRCh38
rs1011669852 Chr.3: 43079529-43079529 on Build GRCh38
rs761474801 Chr.3: 43079730-43079730 on Build GRCh38
rs78518984 Chr.3: 43079537-43079537 on Build GRCh38
rs940678695 Chr.3: 43079523-43079523 on Build GRCh38
rs367979577 Chr.3: 43079640-43079640 on Build GRCh38
rs768219554 Chr.3: 43079710-43079710 on Build GRCh38
rs371375820 Chr.3: 43079707-43079707 on Build GRCh38
rs367621319 Chr.3: 43079693-43079693 on Build GRCh38
rs577978628 Chr.3: 43079610-43079610 on Build GRCh38
rs553792563 Chr.3: 43079531-43079531 on Build GRCh38
rs751429504 Chr.3: 43079667-43079667 on Build GRCh38
rs376572602 Chr.3: 43079677-43079677 on Build GRCh38
rs943093393 Chr.3: 43079622-43079622 on Build GRCh38
rs899215190 Chr.3: 43079548-43079548 on Build GRCh38
rs374779130 Chr.3: 43079685-43079685 on Build GRCh38
rs184464357 Chr.3: 43079583-43079583 on Build GRCh38
rs181123469 Chr.3: 43079550-43079550 on Build GRCh38
rs371760723 Chr.3: 43079617-43079617 on Build GRCh38
rs1011553364 Chr.3: 43079684-43079684 on Build GRCh38
rs554862767 Chr.3: 43079580-43079580 on Build GRCh38
rs531558559 Chr.3: 43079732-43079732 on Build GRCh38
rs148890110 Chr.3: 43079692-43079692 on Build GRCh38
rs775648533 Chr.3: 43079606-43079606 on Build GRCh38
rs1024541848 Chr.3: 43079589-43079589 on Build GRCh38
rs75583919 Chr.3: 43079574-43079574 on Build GRCh38
rs779454247 Chr.3: 43079688-43079688 on Build GRCh38
rs750167854 Chr.3: 43079650-43079650 on Build GRCh38
rs770173778 Chr.3: 43079597-43079597 on Build GRCh38
rs147357138 Chr.3: 43079698-43079698 on Build GRCh38
rs758358962 Chr.3: 43079665-43079665 on Build GRCh38
rs774558057 Chr.3: 43079694-43079694 on Build GRCh38
rs746322589 Chr.3: 43079587-43079587 on Build GRCh38
rs111777493 Chr.3: 43079687-43079687 on Build GRCh38
rs771465179 Chr.3: 43079540-43079540 on Build GRCh38
rs747471745 Chr.3: 43079708-43079708 on Build GRCh38
rs776064566 Chr.3: 43079717-43079717 on Build GRCh38
rs761391441 Chr.3: 43079648-43079648 on Build GRCh38

View Full Product Details