Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 122117937-122118200 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD86
Entrez Gene ID
942
Gene Name
CD86 molecule
Gene Aliases
B7-2, B7.2, B70, CD28LG2, LAB72
UniGene
Hs.171182
Gene Chromosome Location
Chr.3: 122055362-122121143 on Build GRCh38
Transcripts
NM_001206924.1 NM_001206925.1 NM_006889.4 NM_175862.4 NM_176892.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4960284 c.862G>A p.E288K Chr.3: 122118062-122118062 on Build GRCh38
6468364 c.860T>A p.M287K Chr.3: 122118060-122118060 on Build GRCh38
5918014 c.893G>A p.R298K Chr.3: 122118093-122118093 on Build GRCh38
5894785 c.848-7C>T p.? Chr.3: 122118041-122118041 on Build GRCh38
7360316 c.871G>C p.E291Q Chr.3: 122118071-122118071 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs562651636 Chr.3: 122118124-122118124 on Build GRCh38
rs200541347 Chr.3: 122118044-122118044 on Build GRCh38
rs767575031 Chr.3: 122118040-122118040 on Build GRCh38
rs560824456 Chr.3: 122118037-122118037 on Build GRCh38
rs200147755 Chr.3: 122118121-122118121 on Build GRCh38
rs745697549 Chr.3: 122118144-122118144 on Build GRCh38
rs1021454329 Chr.3: 122117975-122117975 on Build GRCh38
rs780946022 Chr.3: 122118135-122118135 on Build GRCh38
rs10580127 Chr.3: 122118120-122118121 on Build GRCh38
rs776452064 Chr.3: 122118011-122118011 on Build GRCh38
rs202079566 Chr.3: 122118122-122118122 on Build GRCh38
rs201982941 Chr.3: 122118063-122118063 on Build GRCh38
rs200746861 Chr.3: 122118004-122118004 on Build GRCh38
rs373110329 Chr.3: 122118127-122118128 on Build GRCh38
rs576843532 Chr.3: 122118058-122118058 on Build GRCh38
rs201100744 Chr.3: 122118131-122118131 on Build GRCh38
rs766702073 Chr.3: 122118049-122118049 on Build GRCh38
rs752337557 Chr.3: 122118066-122118066 on Build GRCh38
rs908132437 Chr.3: 122118089-122118089 on Build GRCh38
rs970243990 Chr.3: 122117976-122117976 on Build GRCh38
rs761240715 Chr.3: 122118048-122118048 on Build GRCh38
rs764319214 Chr.3: 122118021-122118021 on Build GRCh38
rs182035597 Chr.3: 122118139-122118139 on Build GRCh38
rs376329226 Chr.3: 122118098-122118098 on Build GRCh38
rs781192651 Chr.3: 122118117-122118123 on Build GRCh38
rs770701703 Chr.3: 122117999-122117999 on Build GRCh38
rs747318420 Chr.3: 122118118-122118119 on Build GRCh38
rs769514787 Chr.3: 122118145-122118145 on Build GRCh38
rs932089800 Chr.3: 122118128-122118128 on Build GRCh38
rs754275643 Chr.3: 122118050-122118050 on Build GRCh38
rs942221169 Chr.3: 122118094-122118094 on Build GRCh38
rs185834979 Chr.3: 122118169-122118169 on Build GRCh38
rs774514465 Chr.3: 122118028-122118028 on Build GRCh38
rs764788632 Chr.3: 122118176-122118176 on Build GRCh38
rs763207267 Chr.3: 122118015-122118015 on Build GRCh38
rs1020440980 Chr.3: 122117970-122117970 on Build GRCh38
rs777329139 Chr.3: 122118105-122118105 on Build GRCh38

View Full Product Details