Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 113725724-113725997 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP7
Entrez Gene ID
840
Gene Name
caspase 7
Gene Aliases
CASP-7, CMH-1, ICE-LAP3, LICE2, MCH3
UniGene
Hs.9216
Gene Chromosome Location
Chr.10: 113679162-113730909 on Build GRCh38
Transcripts
NM_001227.4 NM_001267056.1 NM_001267057.1 NM_001267058.1 NM_001320911.1 NM_033338.5 NM_033339.4 NM_033340.3 XM_006718017.3 XM_011540260.1 XM_017016763.1 XM_017016764.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5962127 c.370C>T p.R124C Chr.10: 113725856-113725856 on Build GRCh38
5694033 c.392C>G p.P131R Chr.10: 113725878-113725878 on Build GRCh38
7227010 c.485T>C p.I162T Chr.10: 113725971-113725971 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs564010310 Chr.10: 113725807-113725807 on Build GRCh38
rs914789230 Chr.10: 113725809-113725809 on Build GRCh38
rs1011268234 Chr.10: 113725847-113725847 on Build GRCh38
rs1050795948 Chr.10: 113725873-113725873 on Build GRCh38
rs1006520296 Chr.10: 113725882-113725882 on Build GRCh38
rs1040046578 Chr.10: 113725883-113725883 on Build GRCh38
rs928944934 Chr.10: 113725971-113725971 on Build GRCh38
rs970224081 Chr.10: 113725914-113725914 on Build GRCh38
rs368578854 Chr.10: 113725953-113725953 on Build GRCh38
rs566537939 Chr.10: 113725962-113725962 on Build GRCh38
rs993551417 Chr.10: 113725864-113725864 on Build GRCh38
rs890802380 Chr.10: 113725875-113725875 on Build GRCh38
rs889634960 Chr.10: 113725854-113725854 on Build GRCh38
rs768331096 Chr.10: 113725799-113725799 on Build GRCh38
rs188890742 Chr.10: 113725966-113725966 on Build GRCh38
rs117527259 Chr.10: 113725857-113725857 on Build GRCh38
rs1019534316 Chr.10: 113725856-113725856 on Build GRCh38
rs995814830 Chr.10: 113725943-113725943 on Build GRCh38
rs1026712487 Chr.10: 113725865-113725865 on Build GRCh38
rs185649982 Chr.10: 113725853-113725853 on Build GRCh38
rs570085812 Chr.10: 113725867-113725867 on Build GRCh38
rs774821877 Chr.10: 113725905-113725905 on Build GRCh38
rs1007975700 Chr.10: 113725855-113725855 on Build GRCh38
rs984885439 Chr.10: 113725900-113725900 on Build GRCh38
rs534298448 Chr.10: 113725889-113725889 on Build GRCh38
rs774009382 Chr.10: 113725798-113725799 on Build GRCh38
rs142326048 Chr.10: 113725961-113725961 on Build GRCh38

View Full Product Details