Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 197550552-197550777 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPE1-MOB4
Entrez Gene ID
100529241
Gene Name
HSPE1-MOB4 readthrough
Gene Aliases
HSPE1-PHOCN
UniGene
Hs.633165
Gene Chromosome Location
Chr.2: 197499997-197553699 on Build GRCh38
Transcripts
NM_001202485.1 NM_001100819.2 NM_001204094.1 NM_015387.4 NM_199482.3

Gene Symbol
MOB4
Entrez Gene ID
25843
Gene Name
MOB family member 4, phocein
Gene Aliases
2C4D, CGI-95, MOB1, MOB3, MOBKL3, PHOCN, PREI3
UniGene
Hs.633165
Gene Chromosome Location
Chr.2: 197515571-197553699 on Build GRCh38
Transcripts
NM_001202485.1 NM_001100819.2 NM_001204094.1 NM_015387.4 NM_199482.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1637439 c.615C>T p.N205N Chr.2: 197550583-197550583 on Build GRCh38
2905814 c.659C>G p.S220C Chr.2: 197550627-197550627 on Build GRCh38
1614102 c.641A>C p.E214A Chr.2: 197550609-197550609 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1008188593 Chr.2: 197550681-197550681 on Build GRCh38
rs767306009 Chr.2: 197550676-197550676 on Build GRCh38
rs768471095 Chr.2: 197550638-197550638 on Build GRCh38
rs775280983 Chr.2: 197550631-197550631 on Build GRCh38
rs975457026 Chr.2: 197550716-197550716 on Build GRCh38
rs139399234 Chr.2: 197550634-197550634 on Build GRCh38
rs774191648 Chr.2: 197550645-197550645 on Build GRCh38
rs375483019 Chr.2: 197550695-197550695 on Build GRCh38
rs761839479 Chr.2: 197550673-197550673 on Build GRCh38
rs766654247 Chr.2: 197550688-197550688 on Build GRCh38
rs769578142 Chr.2: 197550627-197550627 on Build GRCh38
rs541674711 Chr.2: 197550590-197550590 on Build GRCh38
rs777115834 Chr.2: 197550690-197550690 on Build GRCh38
rs544029948 Chr.2: 197550734-197550734 on Build GRCh38
rs773290934 Chr.2: 197550677-197550677 on Build GRCh38
rs145276780 Chr.2: 197550687-197550687 on Build GRCh38
rs1032020620 Chr.2: 197550655-197550656 on Build GRCh38

View Full Product Details