Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50255666-50255897 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAPK12
Entrez Gene ID
6300
Gene Name
mitogen-activated protein kinase 12
Gene Aliases
ERK-6, ERK3, ERK6, MAPK 12, P38GAMMA, PRKM12, SAPK-3, SAPK3
UniGene
Hs.432642
Gene Chromosome Location
Chr.22: 50252902-50261759 on Build GRCh38
Transcripts
NM_001303252.1 NM_002969.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7261216 c.661+1G>A p.? Chr.22: 50255809-50255809 on Build GRCh38
4387097 c.664C>T p.L222L Chr.22: 50255692-50255692 on Build GRCh38
1417057 c.663C>T p.H221H Chr.22: 50255693-50255693 on Build GRCh38
3759291 c.633T>C p.S211S Chr.22: 50255868-50255868 on Build GRCh38
5456970 c.662-4G>A p.? Chr.22: 50255698-50255698 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148359556 Chr.22: 50255810-50255810 on Build GRCh38
rs369946888 Chr.22: 50255700-50255700 on Build GRCh38
rs781780584 Chr.22: 50255776-50255776 on Build GRCh38
rs775369658 Chr.22: 50255725-50255725 on Build GRCh38
rs141556582 Chr.22: 50255850-50255850 on Build GRCh38
rs753169938 Chr.22: 50255829-50255829 on Build GRCh38
rs886409501 Chr.22: 50255863-50255863 on Build GRCh38
rs1004926265 Chr.22: 50255865-50255865 on Build GRCh38
rs751107042 Chr.22: 50255816-50255816 on Build GRCh38
rs778005801 Chr.22: 50255779-50255779 on Build GRCh38
rs765955866 Chr.22: 50255745-50255745 on Build GRCh38
rs768578561 Chr.22: 50255796-50255796 on Build GRCh38
rs967707656 Chr.22: 50255710-50255710 on Build GRCh38
rs774392515 Chr.22: 50255789-50255789 on Build GRCh38
rs749506892 Chr.22: 50255782-50255782 on Build GRCh38
rs200701475 Chr.22: 50255702-50255702 on Build GRCh38
rs760575782 Chr.22: 50255727-50255727 on Build GRCh38
rs960555806 Chr.22: 50255748-50255748 on Build GRCh38
rs542587121 Chr.22: 50255752-50255752 on Build GRCh38
rs372241388 Chr.22: 50255720-50255720 on Build GRCh38
rs372735885 Chr.22: 50255807-50255807 on Build GRCh38
rs1004916453 Chr.22: 50255695-50255695 on Build GRCh38
rs367769776 Chr.22: 50255871-50255871 on Build GRCh38
rs748476363 Chr.22: 50255777-50255777 on Build GRCh38
rs377211551 Chr.22: 50255830-50255830 on Build GRCh38
rs762907698 Chr.22: 50255867-50255867 on Build GRCh38
rs771024044 Chr.22: 50255722-50255722 on Build GRCh38
rs943591566 Chr.22: 50255795-50255795 on Build GRCh38
rs768008775 Chr.22: 50255827-50255827 on Build GRCh38
rs376320200 Chr.22: 50255778-50255778 on Build GRCh38
rs777972164 Chr.22: 50255843-50255843 on Build GRCh38
rs532716803 Chr.22: 50255874-50255874 on Build GRCh38
rs375314561 Chr.22: 50255689-50255689 on Build GRCh38
rs781305012 Chr.22: 50255692-50255692 on Build GRCh38
rs200047423 Chr.22: 50255822-50255822 on Build GRCh38
rs766188788 Chr.22: 50255814-50255814 on Build GRCh38
rs1012182941 Chr.22: 50255701-50255701 on Build GRCh38
rs774158025 Chr.22: 50255870-50255870 on Build GRCh38
rs373061117 Chr.22: 50255721-50255721 on Build GRCh38
rs977801490 Chr.22: 50255806-50255806 on Build GRCh38
rs1002212449 Chr.22: 50255726-50255726 on Build GRCh38
rs1129880 Chr.22: 50255868-50255868 on Build GRCh38
rs991862197 Chr.22: 50255750-50255750 on Build GRCh38
rs761582494 Chr.22: 50255731-50255731 on Build GRCh38
rs745760808 Chr.22: 50255736-50255737 on Build GRCh38
rs374984773 Chr.22: 50255860-50255860 on Build GRCh38
rs370098873 Chr.22: 50255855-50255855 on Build GRCh38
rs757341538 Chr.22: 50255737-50255737 on Build GRCh38
rs764069068 Chr.22: 50255729-50255729 on Build GRCh38
rs771187307 Chr.22: 50255785-50255785 on Build GRCh38
rs779174381 Chr.22: 50255851-50255851 on Build GRCh38
rs766968717 Chr.22: 50255876-50255876 on Build GRCh38
rs746007209 Chr.22: 50255790-50255790 on Build GRCh38
rs772869234 Chr.22: 50255861-50255861 on Build GRCh38
rs772160186 Chr.22: 50255859-50255859 on Build GRCh38
rs955328658 Chr.22: 50255775-50255775 on Build GRCh38
rs755524400 Chr.22: 50255774-50255774 on Build GRCh38
rs369814739 Chr.22: 50255797-50255797 on Build GRCh38
rs761618832 Chr.22: 50255798-50255798 on Build GRCh38
rs565524902 Chr.22: 50255809-50255809 on Build GRCh38
rs751160165 Chr.22: 50255770-50255770 on Build GRCh38
rs754282586 Chr.22: 50255849-50255849 on Build GRCh38
rs757940462 Chr.22: 50255740-50255740 on Build GRCh38
rs35396905 Chr.22: 50255813-50255813 on Build GRCh38
rs750065014 Chr.22: 50255739-50255739 on Build GRCh38

View Full Product Details