Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 19342777-19343028 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAU2
Entrez Gene ID
23383
Gene Name
MAU2 sister chromatid cohesion factor
Gene Aliases
KIAA0892, MAU2L, SCC4, mau-2
UniGene
Hs.654939
Gene Chromosome Location
Chr.19: 19320686-19358754 on Build GRCh38
Transcripts
NM_015329.3 XM_005259837.3 XM_005259838.3 XM_006722711.3 XM_011527844.2 XM_017026539.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6524803 c.918G>A p.L306L Chr.19: 19342811-19342811 on Build GRCh38
4821215 c.921G>A p.E307E Chr.19: 19342814-19342814 on Build GRCh38
6150004 c.930G>C p.Q310H Chr.19: 19342823-19342823 on Build GRCh38
4075926 c.970A>G p.K324E Chr.19: 19342863-19342863 on Build GRCh38
6150005 c.906G>T p.Q302H Chr.19: 19342799-19342799 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs754845394 Chr.19: 19342904-19342904 on Build GRCh38
rs759929588 Chr.19: 19342875-19342875 on Build GRCh38
rs753852057 Chr.19: 19342903-19342903 on Build GRCh38
rs2301670 Chr.19: 19342913-19342913 on Build GRCh38
rs989874863 Chr.19: 19342966-19342966 on Build GRCh38
rs746638006 Chr.19: 19342919-19342919 on Build GRCh38
rs375842161 Chr.19: 19342814-19342814 on Build GRCh38
rs776591469 Chr.19: 19342929-19342929 on Build GRCh38
rs1032039137 Chr.19: 19342965-19342965 on Build GRCh38
rs1006277160 Chr.19: 19342907-19342907 on Build GRCh38
rs924263091 Chr.19: 19342943-19342943 on Build GRCh38
rs574481366 Chr.19: 19342808-19342808 on Build GRCh38
rs770335527 Chr.19: 19342832-19342832 on Build GRCh38
rs543012604 Chr.19: 19342890-19342890 on Build GRCh38
rs772739904 Chr.19: 19342817-19342817 on Build GRCh38
rs113653692 Chr.19: 19342901-19342901 on Build GRCh38
rs773253002 Chr.19: 19342820-19342820 on Build GRCh38
rs996757621 Chr.19: 19342914-19342914 on Build GRCh38
rs1018417261 Chr.19: 19342989-19342989 on Build GRCh38
rs955707823 Chr.19: 19342958-19342958 on Build GRCh38
rs375089789 Chr.19: 19342912-19342912 on Build GRCh38
rs1006644170 Chr.19: 19342988-19342988 on Build GRCh38
rs751706542 Chr.19: 19342892-19342892 on Build GRCh38
rs954941253 Chr.19: 19342928-19342928 on Build GRCh38
rs777263674 Chr.19: 19342853-19342853 on Build GRCh38
rs2965192 Chr.19: 19342933-19342933 on Build GRCh38
rs371904156 Chr.19: 19342802-19342802 on Build GRCh38
rs192329016 Chr.19: 19342971-19342971 on Build GRCh38
rs999439302 Chr.19: 19342964-19342964 on Build GRCh38
rs886469961 Chr.19: 19342905-19342905 on Build GRCh38
rs368910090 Chr.19: 19342881-19342881 on Build GRCh38
rs909203486 Chr.19: 19342900-19342900 on Build GRCh38
rs985603903 Chr.19: 19342799-19342799 on Build GRCh38
rs767485958 Chr.19: 19342902-19342902 on Build GRCh38
rs775951559 Chr.19: 19342882-19342882 on Build GRCh38
rs914313684 Chr.19: 19342970-19342970 on Build GRCh38
rs758050254 Chr.19: 19342916-19342916 on Build GRCh38
rs559107525 Chr.19: 19342917-19342917 on Build GRCh38
rs867631986 Chr.19: 19342804-19342804 on Build GRCh38

View Full Product Details