Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 32659202-32659439 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYO1D
Entrez Gene ID
4642
Gene Name
myosin ID
Gene Aliases
PPP1R108, myr4
UniGene
Hs.602063
Gene Chromosome Location
Chr.17: 32492522-32877173 on Build GRCh38
Transcripts
NM_001303279.1 NM_015194.2 XM_017024685.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3180483 c.2201A>G p.Y734C Chr.17: 32659259-32659259 on Build GRCh38
6314342 c.2241C>A p.F747L Chr.17: 32659219-32659219 on Build GRCh38
4065422 c.2162C>T p.T721I Chr.17: 32659298-32659298 on Build GRCh38
4065423 c.2149C>T p.R717W Chr.17: 32659311-32659311 on Build GRCh38
705597 c.2129G>A p.R710Q Chr.17: 32659331-32659331 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766821443 Chr.17: 32659297-32659297 on Build GRCh38
rs757145678 Chr.17: 32659247-32659247 on Build GRCh38
rs773557822 Chr.17: 32659290-32659290 on Build GRCh38
rs1010507339 Chr.17: 32659415-32659415 on Build GRCh38
rs745868213 Chr.17: 32659259-32659259 on Build GRCh38
rs765573290 Chr.17: 32659243-32659243 on Build GRCh38
rs768228880 Chr.17: 32659268-32659268 on Build GRCh38
rs753339390 Chr.17: 32659354-32659354 on Build GRCh38
rs767385284 Chr.17: 32659317-32659317 on Build GRCh38
rs759695684 Chr.17: 32659308-32659308 on Build GRCh38
rs956655723 Chr.17: 32659322-32659322 on Build GRCh38
rs76649008 Chr.17: 32659414-32659414 on Build GRCh38
rs935538362 Chr.17: 32659373-32659373 on Build GRCh38
rs771649905 Chr.17: 32659377-32659377 on Build GRCh38
rs748495676 Chr.17: 32659283-32659283 on Build GRCh38
rs772533161 Chr.17: 32659284-32659284 on Build GRCh38
rs778511627 Chr.17: 32659367-32659367 on Build GRCh38
rs762851105 Chr.17: 32659263-32659263 on Build GRCh38
rs753999303 Chr.17: 32659232-32659232 on Build GRCh38
rs753028339 Chr.17: 32659316-32659316 on Build GRCh38
rs756021240 Chr.17: 32659331-32659331 on Build GRCh38
rs368367793 Chr.17: 32659355-32659355 on Build GRCh38
rs760646890 Chr.17: 32659226-32659226 on Build GRCh38
rs979659872 Chr.17: 32659301-32659301 on Build GRCh38
rs372450798 Chr.17: 32659368-32659368 on Build GRCh38
rs776798770 Chr.17: 32659298-32659298 on Build GRCh38
rs770640985 Chr.17: 32659282-32659282 on Build GRCh38
rs761200190 Chr.17: 32659293-32659293 on Build GRCh38
rs1052624478 Chr.17: 32659407-32659407 on Build GRCh38
rs780980753 Chr.17: 32659254-32659254 on Build GRCh38
rs779971386 Chr.17: 32659350-32659350 on Build GRCh38
rs755549663 Chr.17: 32659262-32659262 on Build GRCh38
rs139887773 Chr.17: 32659325-32659325 on Build GRCh38
rs766516579 Chr.17: 32659231-32659231 on Build GRCh38
rs765554830 Chr.17: 32659311-32659311 on Build GRCh38
rs747351334 Chr.17: 32659385-32659385 on Build GRCh38
rs751335872 Chr.17: 32659246-32659246 on Build GRCh38
rs1031071901 Chr.17: 32659391-32659391 on Build GRCh38
rs368852206 Chr.17: 32659378-32659378 on Build GRCh38
rs202148996 Chr.17: 32659236-32659236 on Build GRCh38
rs774055354 Chr.17: 32659281-32659281 on Build GRCh38
rs748810199 Chr.17: 32659266-32659266 on Build GRCh38

View Full Product Details