Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 62369936-62370203 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FEZF2
Entrez Gene ID
55079
Gene Name
FEZ family zinc finger 2
Gene Aliases
FEZ, FEZL, FKSG36, TOF, ZFP312, ZNF312
UniGene
Hs.241523
Gene Chromosome Location
Chr.3: 62369672-62373515 on Build GRCh38
Transcripts
NM_018008.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3824510 c.1291G>C p.D431H Chr.3: 62370172-62370172 on Build GRCh38
1425011 c.1344_1345insC p.S449fs*7 Chr.3: 62370118-62370119 on Build GRCh38
4894425 c.1354G>A p.D452N Chr.3: 62370109-62370109 on Build GRCh38
7168253 c.1282A>G p.R428G Chr.3: 62370181-62370181 on Build GRCh38
3824509 c.1379G>A p.*460* Chr.3: 62370084-62370084 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs994786786 Chr.3: 62370106-62370106 on Build GRCh38
rs759339926 Chr.3: 62370135-62370135 on Build GRCh38
rs765210748 Chr.3: 62370068-62370068 on Build GRCh38
rs186486069 Chr.3: 62370074-62370074 on Build GRCh38
rs774466699 Chr.3: 62370112-62370112 on Build GRCh38
rs201456285 Chr.3: 62370122-62370122 on Build GRCh38
rs370815560 Chr.3: 62370067-62370067 on Build GRCh38
rs74651832 Chr.3: 62370002-62370002 on Build GRCh38
rs751079634 Chr.3: 62370158-62370158 on Build GRCh38
rs770155183 Chr.3: 62370102-62370102 on Build GRCh38
rs767681739 Chr.3: 62370047-62370047 on Build GRCh38
rs541295113 Chr.3: 62370115-62370115 on Build GRCh38
rs201147553 Chr.3: 62370139-62370139 on Build GRCh38
rs746214379 Chr.3: 62370089-62370089 on Build GRCh38
rs751887804 Chr.3: 62370065-62370066 on Build GRCh38
rs1012312280 Chr.3: 62370050-62370050 on Build GRCh38
rs373008990 Chr.3: 62370072-62370072 on Build GRCh38
rs999926126 Chr.3: 62369988-62369988 on Build GRCh38
rs369791744 Chr.3: 62370051-62370051 on Build GRCh38
rs902552065 Chr.3: 62369962-62369962 on Build GRCh38
rs780415973 Chr.3: 62370053-62370053 on Build GRCh38
rs76277372 Chr.3: 62370181-62370181 on Build GRCh38
rs753663399 Chr.3: 62370052-62370052 on Build GRCh38
rs563598995 Chr.3: 62370155-62370155 on Build GRCh38
rs749823763 Chr.3: 62370104-62370104 on Build GRCh38
rs751084158 Chr.3: 62370075-62370075 on Build GRCh38
rs944463706 Chr.3: 62370094-62370094 on Build GRCh38
rs578220193 Chr.3: 62370137-62370137 on Build GRCh38
rs397700951 Chr.3: 62370000-62370001 on Build GRCh38
rs757003054 Chr.3: 62370038-62370039 on Build GRCh38
rs759462594 Chr.3: 62370066-62370066 on Build GRCh38
rs576676584 Chr.3: 62370131-62370131 on Build GRCh38
rs768554364 Chr.3: 62370116-62370116 on Build GRCh38
rs756771992 Chr.3: 62370176-62370176 on Build GRCh38
rs190426453 Chr.3: 62370146-62370146 on Build GRCh38
rs537410902 Chr.3: 62369964-62369964 on Build GRCh38
rs752687331 Chr.3: 62370071-62370071 on Build GRCh38
rs762117256 Chr.3: 62370044-62370044 on Build GRCh38
rs780770480 Chr.3: 62370081-62370081 on Build GRCh38
rs3215030 Chr.3: 62369991-62369992 on Build GRCh38
rs780468998 Chr.3: 62370103-62370103 on Build GRCh38
rs555922430 Chr.3: 62370048-62370048 on Build GRCh38
rs768671004 Chr.3: 62370107-62370107 on Build GRCh38
rs774394149 Chr.3: 62370036-62370036 on Build GRCh38
rs1056434252 Chr.3: 62369990-62369990 on Build GRCh38

View Full Product Details