Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 149970626-149970863 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PFN2
Entrez Gene ID
5217
Gene Name
profilin 2
Gene Aliases
D3S1319E, PFL
UniGene
Hs.91747
Gene Chromosome Location
Chr.3: 149964904-149970954 on Build GRCh38
Transcripts
NM_002628.4 NM_053024.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6394356 c.15G>T p.Q5H Chr.3: 149970842-149970842 on Build GRCh38
6544942 c.23T>A p.V8E Chr.3: 149970834-149970834 on Build GRCh38
6394357 c.15G>T p.Q5H Chr.3: 149970842-149970842 on Build GRCh38
6544943 c.23T>A p.V8E Chr.3: 149970834-149970834 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs888617526 Chr.3: 149970750-149970750 on Build GRCh38
rs902955636 Chr.3: 149970658-149970658 on Build GRCh38
rs773628641 Chr.3: 149970700-149970700 on Build GRCh38
rs926431537 Chr.3: 149970687-149970687 on Build GRCh38
rs771635643 Chr.3: 149970711-149970711 on Build GRCh38
rs188851441 Chr.3: 149970752-149970752 on Build GRCh38
rs1050745451 Chr.3: 149970705-149970705 on Build GRCh38
rs763608984 Chr.3: 149970812-149970812 on Build GRCh38
rs930243825 Chr.3: 149970718-149970718 on Build GRCh38
rs760132081 Chr.3: 149970794-149970794 on Build GRCh38
rs899944102 Chr.3: 149970782-149970782 on Build GRCh38
rs370218022 Chr.3: 149970694-149970694 on Build GRCh38
rs540901618 Chr.3: 149970680-149970680 on Build GRCh38
rs1029609340 Chr.3: 149970719-149970719 on Build GRCh38
rs980929836 Chr.3: 149970653-149970653 on Build GRCh38
rs11559019 Chr.3: 149970770-149970770 on Build GRCh38
rs11559018 Chr.3: 149970826-149970826 on Build GRCh38
rs955365156 Chr.3: 149970747-149970747 on Build GRCh38
rs17851601 Chr.3: 149970749-149970749 on Build GRCh38
rs1032643185 Chr.3: 149970708-149970708 on Build GRCh38
rs998574661 Chr.3: 149970827-149970827 on Build GRCh38
rs1040184893 Chr.3: 149970777-149970777 on Build GRCh38
rs1041837055 Chr.3: 149970646-149970646 on Build GRCh38
rs763287695 Chr.3: 149970706-149970706 on Build GRCh38

View Full Product Details